EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Determination of levamisole and tetramisole in seized cocaine samples by enantioselective high-performance liquid chromatography and circular dichroism detection.
PMID 25124228
Abstract
Levamisole, an anthelmintic drug, has been increasingly employed as an adulterant of illicit street cocaine over the last decade; recently, the use of tetramisole, the racemic mixture of levamisole and its enantiomer dexamisole, was also occasionally observed. A new enantioselective high-performance liquid chromatography (HPLC) method, performed on cellulose tris(3,5-dimethylphenylcarbamate) chiral stationary phases in normal-phase mode, was validated to determine the enantiomeric composition of tetramisole enantiomers in seized cocaine samples. Furthermore, the hyphenation of the validated HPLC method with a circular dichroism (CD) detection system allowed the direct determination of elution order and a selective monitoring of levamisole and dexamisole in the presence of possible interferences. The method was applied to the identification and quantitation of the two enantiomers of tetramisole in seized street cocaine samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1501
Benzocaine
C9H11NO2
06952
Benzocaine, tested according to Ph.Eur.
C9H11NO2
PHR1158
Benzocaine, Pharmaceutical Secondary Standard; Certified Reference Material
C9H11NO2
B0600000
Benzocaine, European Pharmacopoeia (EP) Reference Standard
C9H11NO2
1054000
Benzocaine, United States Pharmacopeia (USP) Reference Standard
C9H11NO2
D0806
Diethylamine, BioXtra
C4H11N
31730
Diethylamine, puriss. p.a., ≥99.5% (GC)
C4H11N
386456
Diethylamine, purified by redistillation, 99.5%
C4H11N
471216
Diethylamine, ≥99.5%
C4H11N
112909
Ethyl 4-aminobenzoate, 98%
C9H11NO2
77440
Phenacetin, ≥98.0% (HPLC)
C10H13NO2
235830
Phenacetin, 97%
C10H13NO2
PHR1249
Phenacetin
C10H13NO2
PHR1094
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
C10H13NO2
1514008
Phenacetin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
C10H13NO2
P9879
Procaine hydrochloride, ≥97%
C13H20N2O2 · HCl
46608
Procaine hydrochloride, VETRANAL, analytical standard
C13H20N2O2 · HCl
1564006
Procaine hydrochloride, United States Pharmacopeia (USP) Reference Standard
C13H20N2O2 · HCl
PHR1161
Procaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C13H20N2O2 · HCl
P3100000
Procaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C13H20N2O2 · HCl