EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Evaluation of sphingomyelin, cholester, and phosphatidylcholine-based immobilized artificial membrane liquid chromatography to predict drug penetration across the blood-brain barrier.
PMID 25124450
Abstract
Over the past decades, several in vitro methods have been tested for their ability to predict drug penetration across the blood-brain barrier. So far, in high-performance liquid chromatography, most attention has been paid to micellar liquid chromatography and immobilized artificial membrane (IAM) LC. IAMLC has been described as a viable approach, since the stationary phase emulates the lipid environment of a cell membrane. However, research in IAMLC has almost exclusively been limited to phosphatidylcholine (PC)-based stationary phases, even though PC is only one of the lipids present in cell membranes. In this article, sphingomyelin and cholester stationary phases have been tested for the first time towards their ability to predict drug penetration across the blood-brain barrier. Upon comparison with the PC stationary phase, the sphingomyelin- and cholester-based columns depict similar predictive performance. Combining data from the different stationary phases did not lead to improvements of the models.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56931
2,6-Diisopropylphenol, analytical standard
C12H18O
A55306
2-(2-Pyridyl)ethylamine, 95%
C7H10N2
B4388
2-Bromo-2-chloro-1,1,1-trifluoroethane, ≥99%
C2HBrClF3
P6273
2-Propylpentanoic acid
C8H16O2
A2169
3′-Azido-3′-deoxythymidine, ≥98% (HPLC)
C10H13N5O4
A7302
4-Acetamidophenol, 98%
C8H9NO2
D8015
4-Dimethylaminoantipyrine, reactive nitrogen species scavenger
C13H17N3O
D4007
5,5-Diphenylhydantoin, ≥99%
C15H12N2O2
1003009
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
C8H9NO2
PHR1005
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
C8H9NO2
A7655
Atenolol, ≥98% (TLC), powder
C14H22N2O3
74827
Atenolol, analytical reference material
C14H22N2O3
1044403
Atenolol, United States Pharmacopeia (USP) Reference Standard
C14H22N2O3
A1340000
Atenolol, European Pharmacopoeia (EP) Reference Standard
C14H22N2O3
12540
Benzene, analytical standard
C6H6
401765
Benzene, anhydrous, 99.8%
C6H6
154628
Benzene, ACS spectrophotometric grade, ≥99%
C6H6
676985
Benzene, ACS reagent, ≥99.0%
C6H6
12553
Benzene, puriss., absolute, over molecular sieve (H2O ≤0.005%), ≥99.5% (GC)
C6H6
12560
Benzene, purum, ≥99.0% (GC)
C6H6
03-0873
Benzene, for residue analysis, suitable for 5000 per JIS
C6H6
03-0910
Benzene, for HPLC
C6H6
PHR1310
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
C6H6
606790
Benzene-12C6, 99.9 atom % 12C
12C6H6
C0100000
Caffeine, European Pharmacopoeia (EP) Reference Standard
C8H10N4O2
1085003
Caffeine, United States Pharmacopeia (USP) Reference Standard
C8H10N4O2
PHR1009
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10N4O2
56396
Caffeine, certified reference material, TraceCERT®
C8H10N4O2
Y0000787
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
C8H10N4O2
1086006
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
C8H10N4O2