EMAIL THIS PAGE TO A FRIEND
Pigment cell & melanoma research
ADAM10 correlates with uveal melanoma metastasis and promotes in vitro invasion.
PMID 25124714
Abstract
Uveal melanoma (UM) is a rare ocular tumor that may lead to deadly metastases in 50% of patients. A disintegrin and metalloproteinase (ADAM)10, ADAM17, and the HGF-receptor c-Met support invasiveness in different tumors. Here, we report that high ADAM10, MET, and, to a lesser extent, ADAM17 gene expression correlates with poor progression-free survival in UM patients (hazard ratio 2.7, 2.6, and 1.9, respectively). About 60% of primary UM expresses c-Met and/or ADAM10 proteins. Four UM cell lines display high levels of ADAM10 and ADAM17, which constitutively cleave c-Met, inducing the release of soluble c-Met. ADAM10/17 pharmacological inhibition or gene silencing reduces c-Met shedding, but has limited impact on surface c-Met, which is overexpressed. Importantly, ADAM10 silencing inhibits UM cell invasion driven by FCS or HGF, while ADAM17 silencing has a limited effect. Altogether our data indicate that ADAM10 has a pro-invasive role and may contribute to UM progression.