EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Simultaneous quantification of 5-hydroxyindoleacetic acid and 5-hydroxytryptamine by capillary electrophoresis with quantum dot and horseradish peroxidase enhanced chemiluminescence detection.
PMID 25125395
Abstract
A capillary electrophoresis (CE) with chemiluminescence (CL) detection method was developed for the simultaneous quantification of 5-hydroxyindoleacetic acid (5-HIAA) and 5-hydroxytryptamine (5-HT). In this method, CdTe quantum dot (QD) and horseradish peroxidase (HRP) were used as enhancing reagents to co-catalyze the post-column CL reaction between luminol and hydrogen peroxide, achieving highly efficient CL emission. 5-HIAA and 5-HT inhibit the CL emission resulting to the formation of negative peaks in electropherogram. The degree of CL suppression is proportional to the concentration of 5-HT and 5-HIAA. The linear ranges for the determination of 5-HIAA and 5-HT were 2.5×10(-8)-2.5×10(-6) M and 2.5×10(-8)-5.0×10(-6) M with detection limits (signal/noise=3) of 7.0×10(-9) M and 6.0×10(-9) M, respectively. Intraday precision do not exceed 5.0%. The accuracy was confirmed by the recoveries ranged from 98% to 104%. The present method was successfully applied for the quantification of 5-HIAA and 5-HT in human urine. The concentrations of 5-HT and 5-HIAA in human urine were found to be in the range of 0.78-1.2 μM and 3.2-5.1 μM, respectively.