EMAIL THIS PAGE TO A FRIEND
Journal of Alzheimer's disease : JAD
Impaired lysosomal cobalamin transport in Alzheimer's disease.
PMID 25125476
Abstract
Cobalamin (vitamin B12) is required for erythrocyte formation and DNA synthesis and it plays a crucial role in maintaining neurological function. As a coenzyme for methionine synthase and methylmalonyl-CoA mutase, cobalamin utilization depends on its efficient transit through the intracellular lysosomal compartment and subsequent delivery to the cytosol and mitochondria. Lysosomal function deteriorates in Alzheimer's disease (AD). Lysosomal acidification is defective in AD and lysosomal proteolysis is disrupted by AD-related presenilin 1 mutation. In this study, we propose that AD related lysosomal dysfunction may impair lysosomal cobalamin transport. The experiments use in vitro and in vivo models of AD to define how lysosomal dysfunction directly affects cobalamin utilization. SH-SY5Y-AβPP mutant cells were treated with a proteasome inhibitor to induce lysosomal amyloid-β accumulation. We metabolically labeled these cells with [57Co] cobalamin and isolated purified lysosomes, mitochondria, and cytosol fractions. The results indicated that proteasome inhibition was associated with lysosomal amyloid-β accumulation and a doubling of lysosomal [57Co] cobalamin levels. We also used AβPPxPS1 transgenic AD mice that were intraperitoneally injected with [57Co] cobalamin. The amount of [57Co] cobalamin in the major organs of these mice was measured and the subcellular [57Co] cobalamin distribution in the brain was assessed. The results demonstrated that lysosomal [57Co] cobalamin level was significantly increased by 56% in the AβPPxPS1 AD mouse brains as compared to wild type control mice. Together these data provide evidence that lysosomal cobalamin may be impaired in AD in association with amyloid-β accumulation.