EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
A generic approach to post-column refocusing in liquid chromatography.
PMID 25127691
Abstract
To increase detection sensitivity in liquid chromatography, a generic post-column refocusing strategy has been developed to enrich (target) analytes prior to detection. In this strategy, after separation on the analytical column, the analytes are led to a trap column preferably containing a stationary phase with strong retentive properties (e.g. silica C30). They are then eluted using a strong solvent in a backward-elution mode. A first key element of the proposed strategy is that the trapping time should be at least equal to the time the front of the remobilization solvent needs to cover the entire length of the trap column, divided by the ratio of the flow rates used for trapping and remobilization. This condition is independent of the retention properties of the analytes in the trapping and remobilization solvent. Another essential element is the addition of a third solvent (isopropanol in the present case) to the remobilization solvent to overcome viscous-fingering effects caused by the viscosity difference between the trap and the remobilization solvents. The potential of the proposed post-column refocusing strategy is demonstrated for an isocratic separation of KI (t0 marker), an antibiotic (sulfamethazine), and acetophenone as a case study. Using optimized remobilization conditions a maximum signal-enhancement factor of 8 was achieved. Higher enhancement factors using a remobilization solvent with slightly higher elution strength were prohibited by disturbances of the UV background signal.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63634
Acetophenone, TraceCERT®, certified reference material
C8H8O
42163
Acetophenone, analytical standard
C8H8O
00790
Acetophenone, puriss. p.a., ≥99.0% (GC)
C8H8O
A10701
Acetophenone, ReagentPlus®, 99%
C8H8O
W200905
Acetophenone, ≥98%, FG
C8H8O
W200910
Acetophenone, natural, 98%, FG
C8H8O
746428
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
IK
793582
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
IK
60399
Potassium iodide, BioUltra, ≥99.5% (AT)
IK
P2963
Potassium iodide, BioXtra, ≥99.0%
IK
30315
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.5%
IK
P8166
Potassium iodide, plant cell culture tested
IK
03124
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
IK
60422
Potassium iodide, tested according to Ph.Eur.
IK
429422
Potassium iodide, anhydrous, beads, −10 mesh, 99.998% trace metals basis
IK
204102
Potassium iodide, ≥99.99% trace metals basis
IK
PHR1360
Potassium Iodide, Pharmaceutical Secondary Standard; Certified Reference Material
IK
S1900000
Sulfadimidine, European Pharmacopoeia (EP) Reference Standard
C12H14N4O2S
Y0001495
Sulfadimidine for peak identification, European Pharmacopoeia (EP) Reference Standard
C12H14N4O2S
00189
Sulfadimidine solution, 1 mg/mL in H2O
C12H14N4O2S
S6256
Sulfamethazine, ≥99%
C12H14N4O2S
46802
Sulfamethazine, VETRANAL, analytical standard
C12H14N4O2S