EMAIL THIS PAGE TO A FRIEND
Prostaglandins, leukotrienes, and essential fatty acids
Effects of chronic administration of arachidonic acid on lipid profiles and morphology in the skeletal muscles of aged rats.
PMID 25128088
Abstract
Arachidonic acid (20:4n-6, ARA) is a major component of the cell membrane, whereas ARA-derived eicosanoids are formed when cells are damaged. Aging is associated with an accretion of oxidative stress in skeletal muscles. In this study, we examined the effects of chronic administration (13 weeks) of ARA (240 mg/kg/day) on fatty acid composition, antioxidative status, and morphology of slow (soleus muscles) and fast (extensor digitorum longus muscles; EDL)-twitch muscles in aged rats (21 months old). The level of reactive oxygen species was higher in the EDL of ARA-administered rats than in that of control rats. ARA administration decreased the muscle cell volumes and increased the number of slow myosin heavy chain (MHC)-positive cells in the EDL. The relative content of MHC2X was increased whereas the relative content of MHC2B was decreased in the EDL of ARA-administered rats. These results suggest that ARA deposition in the fast-twitch muscle of aged rats reduced cell volume with an increase in oxidative stress.