EMAIL THIS PAGE TO A FRIEND
Cancer letters
Lysyl oxidase-like 2 (LOXL2) from stromal fibroblasts stimulates the progression of gastric cancer.
PMID 25128648
Abstract
The aim of this study was to clarify the role of fibroblast-derived Lysyl oxidase-like 2 (LOXL2) in the development of gastric cancer. The correlation between the clinicopathological features of 548 primary gastric carcinomas and LOXL2 expression in stromal cells was examined by immunohistochemistry. Two gastric cancer cell lines, OCUM-12 and NUGC-3, and cancer-associated fibroblasts (CAFs) were used in this in vitro study. The effect of fibroblast-derived LOXL2 on the motility of gastric cancer cells was analyzed by using a wound-healing assay, a double-chamber invasion assay, and western blot. LOXL2 expression in stromal cells was significantly associated with tumor invasion depth, lymph node metastasis, lymphatic invasion, venous invasion, and peritoneal dissemination. Multivariable logistic regression analysis revealed that LOXL2 expression in stromal cells could be an independent predictive parameter for the overall survival of patients. CAFs significantly stimulated the migration and invasion of OCUM-12 and NUGC-3 cells. This motility-stimulating ability of CAFs was inhibited by LOXL2 siRNA. Western blot analysis indicated that phosphorylation of focal adhesion kinase (FAK) in cancer cells was increased by the conditioned medium from CAFs, and was decreased by the conditioned medium from LOXL2 siRNA-treated CAFs. LOXL2 expression in stromal cells may be a useful prognostic factor for patients with gastric cancer. Fibroblast-derived LOXL2 may stimulate the motility of gastric cancer cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1100133
Anti-LOXL2 (606-620) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2107068
Anti-LOXL2 antibody produced in rabbit, affinity isolated antibody
SAB4504583
Anti-phospho-FAK (pSer843) antibody produced in rabbit, affinity isolated antibody
SAB4504180
Anti-phospho-FAK (pSer910) antibody produced in rabbit, affinity isolated antibody
F7926 Anti-phospho-FAK (pTyr397) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504181
Anti-phospho-FAK (pTyr397) antibody produced in rabbit, affinity isolated antibody
SAB4504403
Anti-phospho-FAK (pTyr397) antibody produced in rabbit, affinity isolated antibody
F8051 Anti-phospho-FAK (pTyr407) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503913
Anti-phospho-FAK (pTyr407) antibody produced in rabbit, affinity isolated antibody
SAB4503869
Anti-phospho-FAK (pTyr576) antibody produced in rabbit, affinity isolated antibody
F8926 Anti-phospho-FAK (pTyr577) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
F9176 Anti-phospho-FAK (pTyr861) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504404
Anti-phospho-FAK (pTyr861) antibody produced in rabbit, affinity isolated antibody
SAB4504405
Anti-phospho-FAK (pTyr925) antibody produced in rabbit, affinity isolated antibody
SAB4300281
Anti-phospho-PTK2 (pTyr576/pTyr577) antibody produced in rabbit, affinity isolated antibody
SAB4300047
Anti-phospho-PTK2 (pTyr861) antibody produced in rabbit, affinity isolated antibody
SAB4300101
Anti-phospho-PTK2 (pTyr925) antibody produced in rabbit, affinity isolated antibody
WH0004017M1
Monoclonal Anti-LOXL2 antibody produced in mouse, clone 5D2, purified immunoglobulin, buffered aqueous solution