EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
A novel surface-confined glucaminium-based ionic liquid stationary phase for hydrophilic interaction/anion-exchange mixed-mode chromatography.
PMID 25129388
Abstract
Glucaminium-based ionic liquids are a new class of recently developed ionic liquids and prepared by functionalizing the amine group of N-methyl-d-glucamine, which renders them good hydrophilicity due to the presence of the glucose structure and charged quaternary ammonium group. In the present study, a glucaminium-based ionic liquid N,N-diallyl-N-methyl-d-glucaminium bromide was synthesized and subsequently bonded to the surface of 3-mercaptopropyl modified silica materials through "thiol-ene" click chemistry. The obtained stationary phase was characterized by elemental analysis and infrared spectroscopy, and then packed as a HPLC column. A mixture of five nucleosides was used to characterize the separation performance of the obtained column under HILIC mode and the column efficiency was determined with cytidine as the test solute, reaching 80,000plates/m. Then, the retention behavior was evaluated by investigating the effect of various chromatographic factors on retention of different types of solutes, and the results revealed that the developed surface-confined glucaminium-based ionic liquid stationary phase exhibited a hydrophilic interaction/anion-exchange mixed-mode retention mechanism. Finally, two mixtures of nucleotides and flavonoids were separated on the glucaminium-based ionic liquid column, respectively under hydrophilic interaction and hydrophilic interaction/anion-exchange mixed-mode chromatography. In conclusion, the multimodal retention capabilities of the glucaminium-based ionic liquid column could offer a wider range of retention behavior and flexible selectivity toward polar and hydrophilic compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
01897
Adenosine 5′-diphosphate disodium salt, ≥90%
C10H13N5Na2O10P2
A2252
Adenosine 5′-monophosphate monohydrate, from yeast, ≥97%
C10H14N5O7P · H2O
FLAAS
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of ~1 mg ATP
C10H14N5Na2O13P3 · xH2O
A1852
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of 30 mg
C10H14N5Na2O13P3 · xH2O
A7699
Adenosine 5′-triphosphate disodium salt hydrate, BioXtra, ≥99% (HPLC), from microbial
C10H14N5Na2O13P3 · xH2O
A26209
Adenosine 5′-triphosphate disodium salt hydrate, 99%
C10H14N5Na2O13P3 · xH2O
A2383
Adenosine 5′-triphosphate disodium salt hydrate, Grade I, ≥99%, from microbial
C10H14N5Na2O13P3 · xH2O
A3377
Adenosine 5′-triphosphate disodium salt hydrate, Grade II, ≥98.5% (HPLC), crystalline, from microbial
C10H14N5Na2O13P3 · xH2O
A6419
Adenosine 5′-triphosphate disodium salt hydrate, microbial, BioReagent, suitable for cell culture, ≥99% (HPLC)
C10H14N5Na2O13P3 · xH2O
C4654
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
C9H13N3O5
C122106
Cytidine, 99%
C9H13N3O5
30270
Cytidine, ≥99.0% (HPLC)
C9H13N3O5
C9755
Cytidine 5′-diphosphate sodium salt hydrate, from yeast, ≥95%
C9H15N3O11P2
C1131
Cytidine 5′-monophosphate, Sigma Grade, ≥99% (HPLC), synthetic, powder
C9H14N3O8P
G7127
Guanosine 5′-diphosphate sodium salt, Type I, ≥96% (HPLC)
C10H15N5O11P2
G8377
Guanosine 5′-monophosphate disodium salt hydrate, from yeast, ≥99%
C10H12N5Na2O8P · xH2O
I4125
Inosine, ≥99% (HPLC)
C10H12N4O5
I2753
Inosine 3′:5′-cyclic monophosphate sodium salt, ≥98%
C10H10N4NaO7P
I4375
Inosine 5′-diphosphate sodium salt, ≥96%
C10H14N4O11P2
PHR1324
Meglumine, Pharmaceutical Secondary Standard; Certified Reference Material
C7H17NO5
66930
N-Methyl-D-glucamine, ReagentPlus®, ≥99.0% (T)
C7H17NO5
Y0001209
N-Methyl-D-glucamine, European Pharmacopoeia (EP) Reference Standard
C7H17NO5
282626
Trimethoxysilane, 95%
C3H10O3Si