EMAIL THIS PAGE TO A FRIEND
Current biology : CB
Endogenous species of mammalian nonmuscle myosin IIA and IIB include activated monomers and heteropolymers.
PMID 25131674
Abstract
Class II myosins generate contractile forces in cells by polymerizing into bipolar filaments and pulling on anchored actin filaments. Nonmuscle myosin II (NMII) plays central roles during cell adhesion, migration, cytokinesis, and tissue morphogenesis. NMII is present in virtually all mammalian cell types as tissue-specific combinations of NMIIA, NMIIB, and NMIIC isoforms. It remains poorly understood how the highly dynamic NMII-actin contractile system begins to assemble at new cellular locations during cell migration and how incorporation of different NMII isoforms into this system is coordinated. Using platinum replica electron microscopy in combination with immunogold labeling, we demonstrate that individual activated (phosphorylated on the regulatory light chain andxa0unfolded) NMIIA and NMIIB molecules represent a functional form of NMII in motile cells and that NMIIA and NMIIB copolymerize into nascent bipolar filaments during contractile system assembly. Using subdiffraction stimulated emission depletion microscopy together with a pharmacological block-and-release approach, we report that NMIIA and NMIIB simultaneously incorporate into the cytoskeleton during initiation of contractile system assembly, whereas the characteristic rearward shift of NMIIB relative to NMIIA is established later in the course of NMII turnover. We show existence of activated NMII monomers in cells, copolymerization of endogenous NMIIA and NMIIB molecules, and contribution of both isoforms, rather than only NMIIA, to early stages of the contractile system assembly. These data change the current paradigms about dynamics and functions of NMII and provide new conceptual insights into the organization and dynamics of the ubiquitous cellular machinery for contraction that acts in multiple cellular contexts.