PloS one
Antioxidant and antiproliferative activities of twenty-four Vitis vinifera grapes.
PMID 25133401
Abstract
Grapes are rich in phytochemicals with many proven health benefits. Phenolic profiles, antioxidant and antiproliferative activities of twenty-four selected Vitis vinifera grape cultivars were investigated in this study. Large ranges of variation were found in these cultivars for the contents of total phenolics (95.3 to 686.5 mg/100 g) and flavonoids (94.7 to 1055 mg/100 g) and antioxidant activities (oxygen radical absorbance capacity 378.7 to 3386.0 mg of Trolox equivalents/100 g and peroxylradical scavenging capacity14.2 to 557 mg of vitamin C equivalents/100 g), cellular antioxidant activities (3.9 to 139.9 µmol of quercetin equivalents/100 g without PBS wash and 1.4 to 95.8 µmol of quercetin equivalents /100 g with PBS wash) and antiproliferative activities (25 to 82% at the concentrations of 100 mg/mL extracts).The total antioxidant activities were significantly correlated with the total phenolics and flavonoids. However, no significant correlations were found between antiproliferative activities and total phenolics or total flavonoids content. Wine grapes and color grapes showed much higher levels of phytochemicals and antioxidant activities than table grapes and green/yellow grapes. Several germplasm accessions with much high contents of phenolics and flavonoids, and total antioxidant activity were identified. These germplasm can be valuable sources of genes for breeding grape cultivars with better nutritional qualities of wine and table grapes in the future.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
03382
Aluminum atomic spectroscopy standard concentrate 10.00 g Al, 10.00 g/L, for 1 l standard solution, analytical standard
AlCl3
563919
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
AlCl3
294713
Aluminum chloride, 99.99% trace metals basis
AlCl3
449598
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
AlCl3
495352
Aluminum chloride, ReagentPlus®, anhydrous, ≥99.9% trace metals basis
AlCl3
06230
Aluminum chloride, purum, anhydrous, ≥98.0% (AT)
AlCl3
06154
Aluminum standard for AAS, analytical standard, traceable to BAM
AlCl3
45374
Chloranil, PESTANAL®, analytical standard
C6Cl4O2
23290
Chloranil, purum, ≥97.0% (AT)
C6Cl4O2
77634
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
C8H8O3
232017
Tetrachloro-1,4-benzoquinone, 99%
C6Cl4O2
V2375
Vanillin
C8H8O3
30304
Vanillin, TraceCERT®, certified reference material
C8H8O3
94752
Vanillin, tested according to Ph.Eur.
C8H8O3
PHR1245
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
C8H8O3
V1104
Vanillin, ReagentPlus®, 99%
C8H8O3
W310700
Vanillin, ≥97%, FCC, FG
C8H8O3
W310727
Vanillin, natural, ≥97%, FCC, FG
C8H8O3
V0050000
Vanillin, European Pharmacopoeia (EP) Reference Standard
C8H8O3
PHR1091
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
C8H8O3
1711009
Vanillin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
C8H8O3