PloS one
Nuclear translocation of hARD1 contributes to proper cell cycle progression.
PMID 25133627
Abstract
Arrest defective 1 (ARD1) is an acetyltransferase that is highly conserved across organisms, from yeasts to humans. The high homology and widespread expression of ARD1 across multiple species and tissues signify that it serves a fundamental role in cells. Human ARD1 (hARD1) has been suggested to be involved in diverse biological processes, and its role in cell proliferation and cancer development has been recently drawing attention. However, the subcellular localization of ARD1 and its relevance to cellular function remain largely unknown. Here, we have demonstrated that hARD1 is imported to the nuclei of proliferating cells, especially during S phase. Nuclear localization signal (NLS)-deleted hARD1 (hARD1ΔN), which can no longer access the nucleus, resulted in cell morphology changes and cellular growth impairment. Notably, hARD1ΔN-expressing cells showed alterations in the cell cycle and the expression levels of cell cycle regulators compared to hARD1 wild-type cells. Furthermore, these effects were rescued when the nuclear import of hARD1 was restored by exogenous NLS. Our results show that hARD1 nuclear translocation mediated by NLS is required for cell cycle progression, thereby contributing to proper cell proliferation.