EMAIL THIS PAGE TO A FRIEND
Leukemia
CD34(+)CD38(-)CD58(-) cells are leukemia-propagating cells in Philadelphia chromosome-positive acute lymphoblastic leukemia.
PMID 25135692
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N9879 NADase from porcine brain, ≥0.007 unit/mg solid