EMAIL THIS PAGE TO A FRIEND
ChemSusChem
Diglycerol-based polyesters: melt polymerization with hydrophobic anhydrides.
PMID 25138308
Abstract
The melt polymerization of diglycerol with bicyclic anhydride monomers derived from a naturally occurring monoterpene provides an avenue for polyesters with a high degree of sustainability. The hydrophobic anhydrides are synthesized at ambient temperature via a solvent-free Diels-Alder reaction of α-phellandrene with maleic anhydride. Subsequent melt polymerizations with tetra-functional diglycerol are effective under a range of [diglycerol]/[anhydride] ratios. The hydrophobicity of α-phellandrene directly impacts the swelling behavior of the resulting polyesters. The low E factors (<2), large amount of bio-based content (>75%), ambient temperature monomer synthesis, and polymer degradability represent key factors in the design of these sustainable polyesters.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
240559
1,4-Butanediol, ReagentPlus®, ≥99%
C4H10O2
493732
1,4-Butanediol, ReagentPlus®, 99%
C4H10O2