EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
TGF-β1 enhances SDF-1-induced migration and tube formation of choroid-retinal endothelial cells by up-regulating CXCR4 and CXCR7 expression.
PMID 25138701
Abstract
Stromal derived factor (SDF)-1 has been confirmed to regulate angiogenesis in choroidal neovascularization formation via its two receptors, CXC chemokine receptors 4 (CXCR4) and 7 (CXCR7). Previous studies found that there is cross-talk between the transforming growth factor beta (TGF-β) and SDF-1 pathways in some types of immune or tumor cells, but much less is known about this interaction in endothelial cells. This study investigated the effects of TGF-β1 on CXCR4 and CXCR7 expression as well as SDF-1-induced migration and tube formation in choroid-retinal endothelial (RF/6A) cells. RF/6A cells were treated with recombinant TGF-β1 at various concentrations and time points. Real-time PCR and Western blotting were used to examine the mRNA and protein levels of CXCR4 and CXCR7. In addition, transwell migration and Matrigel tube formation analyses were performed to investigate the role of TGF-β1 pretreatment in SDF-1-induced RF/6A cell migration and tube formation. Our results showed that treatment with recombinant human TGF-β1 enhanced the CXCR4 and CXCR7 levels in time- and dose-dependent manners. The increased CXCR4 and CXCR7 expression resulted in increased SDF-1-induced RF/6A cell migration and tube formation. In addition, the transcriptional regulation of CXCR4 and CXCR7 by TGF-β1 was found to be mediated by phosphorylation of the extracellular signal-related kinase1/2 pathway. Altogether, these results demonstrate that a cross-talk exists between the TGF-β1 and SDF-1 pathways in choroid-retinal endothelial cells, reflecting a novel molecular mechanism that explains the pro-angiogenic effects of TGF-β1 and possibly provides new perspectives for the treatment of CNV-associated diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S0447
SIS3, ≥98% (HPLC), powder
C28H27N3O3 · HCl