EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Development and validation of a hydrophilic interaction liquid chromatography with tandem mass spectrometry method for the simultaneous detection and quantification of etilefrine and oxilofrine in equine blood plasma and urine.
PMID 25143252
Abstract
A sensitive hydrophilic interaction liquid chromatography coupled with tandem mass spectrometry method was developed and validated for the simultaneous detection and quantification of etilefrine and oxilofrine in equine blood plasma and urine. The method is highly sensitive and specific with good precision and accuracy. In plasma the limit of detection and limit of quantification are 0.03 and 0.1xa0ng/mL, respectively, for both analytes. In urine the limit of detection and limit of quantification are 0.3 and 1xa0ng/mL, respectively, for both analytes. The suitability of the method for doping control analysis in equine species is demonstrated by analyzing postadministration samples collected after a single intravenous administration of 50xa0mg etilefrine to a standardbred mare. Etilefrine was detected up to 120xa0h in urine and up to 48xa0h in plasma. Etilefrine is highly conjugated in equine urine whereas it exists in the free form in equine plasma. Therefore, enzyme hydrolysis prior to sample preparation is recommended for the detection and quantification of etilefrine and oxilofrine in equine urine.