EMAIL THIS PAGE TO A FRIEND
Disease markers
Recipients with in utero induction of tolerance upregulated MHC class I in the engrafted donor skin.
PMID 25143665
Abstract
The alterations in MHC class I expression play a crucial step in immune evasion of cancer or virus-infected cells. This study aimed to examine whether tolerized grafts modified MHC class I expression. FVB/N mice were rendered tolerant of C57BL/6 alloantigens by in utero transplantation of C57BL/6 marrows. Postnatally, engrafted donor skins and leukocytes were examined for their MHC expression by quantitative real-time PCR and flow cytometry. Engrafted donor skins upregulated their MHC class I related gene transcripts after short-term (1~2 weeks) or long-term (>1 month) engraftment. This biological phenomenon was simultaneously associated with upregulation of TAP1 gene transcripts, suggesting an important role of TAP1 in the regulation of MHC class I pathway. The surface MHC class I molecules of H-2K(b) in engrafted donor leukocytes consistently showed overexpression. Conclusively, the induction of allograft tolerance involved biological modifications of donor transplants. The overexpression of MHC class I within engrafted transplants of tolerant mice might be used as the tolerance biomarkers for identifying a state of graft tolerance.