EMAIL THIS PAGE TO A FRIEND
Nature communications
Twist1 induces endothelial differentiation of tumour cells through the Jagged1-KLF4 axis.
PMID 25146389
Abstract
The mechanisms controlling tumour-induced angiogenesis are presently not clear. In principle, angiogenesis can be achieved through the activation of endothelial cells in existing vessels or by transdifferentiation of tumour cells into endothelial cells. However, whether tumour cells can go through a prior epithelial-mesenchymal transition and further differentiate into endothelial cells remains unknown. Here we show that overexpression of Twist1, a transcriptional regulator that induces and promotes cancer metastasis, leads to endothelial differentiation in head and neck cancer (HNC) cells. Induction of Jagged1 expression by Twist1 is essential for Twist1-induced endothelial differentiation. The Jagged1/Notch signalling subsequently activates KLF4, inducing stem-like properties in HNC cells and conferring them with drug resistance. Our results indicate that the Twist1-Jagged1/KLF4 axis is essential both for transdifferentiation of tumour cells into endothelial cells and for chemoresistance acquisition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4200033
Anti-BMI1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA030472
Anti-BMI1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106309
Anti-BMI1 antibody produced in rabbit, affinity isolated antibody
AV38430
Anti-KLF4 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
AV35721
Anti-KLF4 antibody produced in rabbit, IgG fraction of antiserum
HPA002926
Anti-KLF4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1300678
Anti-KLF4 antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB2105308
Anti-KLF4 antibody produced in rabbit, affinity isolated antibody
SAB2701975
Anti-KLF4 antibody produced in rabbit
SAB2106435
Anti-KLF4 antibody produced in rabbit, affinity isolated antibody
AV34213
Anti-PCGF4 antibody produced in rabbit, affinity isolated antibody
SAB5300333
Monoclonal Anti-BMI1 antibody produced in mouse, clone 3E3, ascites fluid
SAB5300069
Monoclonal Anti-KLF4 antibody produced in mouse, clone 1E6, ascites fluid
WH0000648M2
Monoclonal Anti-PCGF4 antibody produced in mouse, clone 4E10-1C5, purified immunoglobulin, buffered aqueous solution
PLA0208
Rabbit anti-BMI1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.