EMAIL THIS PAGE TO A FRIEND
Investigative ophthalmology & visual science
The resolvin D1 analogue controls maturation of dendritic cells and suppresses alloimmunity in corneal transplantation.
PMID 25146982
Abstract
To analyze the effect of a resolvin D1 (RvD1) analogue (RvD1a) on dendritic cell maturation, T-cell sensitization, and allograft rejection in corneal allotransplantation. The receptor expression of RvD1 (ALX/FPR2) on bone marrow-derived dendritic cells (BMDC) was measured using quantitative real-time PCR. We determined BMDC maturation after treatment with RvD1a using ELISA to measure interleukin (IL)-12 protein expression and flow cytometry to assess the expression of CD40, major histocompatibility complex (MHC) II, CD80, and CD86. After corneal transplantation in BALB/c mice, we analyzed T-cell infiltration in the cornea and the draining lymph nodes using flow cytometry. The enzyme-linked immunospot (ELISPOT) assay was used to measure T-cell sensitization via the direct and indirect pathway. Angiogenesis and lymphangiogenesis in the cornea after transplantation were measured using immunohistochemistry. Graft opacity and survival were evaluated by slit lamp biomicroscopy. The receptor for RvD1, lipoxin A4/formyl peptide receptor 2 (ALX/FPR2), was expressed at a significantly lower level on immature than mature dendritic cells (DCs), and RvD1a reduced DC expression of MHC II, CD40, and IL-12 following lipopolysaccharide (LPS) stimulation. Using a murine model of corneal transplantation, RvD1a-treated hosts exhibited significantly reduced allosensitization as demonstrated by decreased frequencies of interferon-gamma-secreting T cells in the draining lymph nodes, and reduced T-cell infiltration into the grafts. Graft survival was significantly enhanced and angiogenesis at the graft site was suppressed in RvD1a-treated hosts compared with vehicle-treated hosts. These results suggest that RvD1 inhibits DC maturation and reduces alloimmune sensitization following transplantation, thereby establishing a novel connection between resolvin D1 and the regulation of DC-mediated, antigen-specific immunity.