EMAIL THIS PAGE TO A FRIEND
Food chemistry
Fractionation and identification of minor and aroma-active constituents in Kangra orthodox black tea.
PMID 25148991
Abstract
The aroma constituents of Kangra orthodox black tea were isolated by simultaneous distillation extraction (SDE), supercritical fluid extraction and beverage method. The aroma-active compounds were identified using gas chromatography-olfactometry-mass spectrometry. Geraniol, linalool, (Z/E)-linalool oxides, (E)-2-hexenal, phytol, β-ionone, hotrienol, methylpyrazine and methyl salicylate were major volatile constituents in all the extracts. Minor volatile compounds in all the extracts were 2-ethyl-5-methylpyrazine, ethylpyrazine, 2-6,10,14-trimethyl-2-pentadecanone, acetylfuran, hexanoic acid, dihydroactinidiolide and (E/Z)-2,6-nonadienal. The concentrated SDE extract was fractionated into acidic, basic, water-soluble and neutral fractions. The neutral fraction was further chromatographed on a packed silica gel column eluted with pentane and diethyl ether to separate minor compounds. The aroma-active compounds identified using gas chromatography-olfactometry-mass spectrometry were 2-amylfuran, (E/Z)-2,6-nonadienal, 1-pentanol, epoxylinalool, (Z)-jasmone, 2-acetylpyrrole, farnesyl acetone, geranyl acetone, cadinol, cubenol and dihydroactinidiolide. AEDA studies showed 2-hexenal, 3-hexenol, ethylpyrazine, (Z/E)-linalool oxides, linalool, (E/Z)-2,6-nonadienal, geraniol, phenylethanol, β-ionone, hotrienol and dihydroactinidiolide to be odour active components.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74856
(−)-Linalool, analytical standard
C10H18O
62139
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
C10H18O
P13800
1-Phenylethanol, 98%
C8H10O
W268518
α-Methylbenzyl alcohol, ≥99%, FCC, FG
C8H10O
268909
cis-3,7-Dimethyl-2,6-octadien-1-ol, 97%
C10H18O
W277002
cis-3,7-Dimethyl-2,6-octadien-1-ol, ≥97%, FCC, FG
C10H18O
48798
Geraniol, analytical standard
C10H18O
163333
Geraniol, 98%
C10H18O
W250708
Geraniol, ≥97%, FCC, FG
C10H18O
W250716
Geraniol, natural, ≥97%, FG
C10H18O
51782
Linalool, analytical standard
C10H18O
W263508
Linalool, ≥97%, FCC, FG
C10H18O
L2602
Linalool, 97%
C10H18O
M6752
Methyl salicylate, ReagentPlus®, ≥99% (GC)
C8H8O3
76631
Methyl salicylate, analytical standard
C8H8O3
M2047
Methyl salicylate, BioXtra
C8H8O3
84332
Methyl salicylate, tested according to Ph.Eur.
C8H8O3
1437450
Methyl salicylate, United States Pharmacopeia (USP) Reference Standard
C8H8O3
W274518
Methyl salicylate, ≥98%, FCC, FG
C8H8O3
PHR1214
Methyl salicylate, Pharmaceutical Secondary Standard; Certified Reference Material
C8H8O3
50949
Nerol, analytical standard
C10H18O
H59605
Nerolidol, 98%, Mixture of cis and trans
C15H26O
W277207
Nerolidol, mixture of cis and trans, ≥97%, stabilized, FG
C15H26O
00459
Nerolidol, mixture of cis and trans, analytical reference material
C15H26O