EMAIL THIS PAGE TO A FRIEND
Acta tropica
Inhibition of metalloproteinase-9 secretion and gene expression by artemisinin derivatives.
PMID 25149353
Abstract
Malaria remains one of the world's most common infectious diseases, being responsible for more deaths than any other communicable disease except tuberculosis. There is strong evidence that tumour necrosis factor α and interleukin-1β are important contributors to the systemic disease caused by the infection with Plasmodium falciparum. Circulating levels of TNFα are increased after infection, as a consequence of stimulation of monocyte-macrophages by infected red blood cells or parasite products, as shown in vitro for the malaria pigment haemozoin. TNFα in turn enhances the synthesis of metalloproteinase-9 in monocytes and macrophages. Metalloproteinase-9 acts on the extracellular matrix but also on non-traditional substrates, including precursors of inflammatory cytokines, which are proteolytically activated and contribute to the amplification of the inflammatory response. The aim of the present work was to establish whether artemisinin and its derivatives artemisone, artesunate and dihydroartemisinin possess immuno-modulatory properties. In particular, it is necessary to evaluate their effects on mRNA levels and secretion of MMP-9 by the human monocytic cell line (THP-1 cells) stimulated by hemozoin or TNFα. 5μM of each derivative, although not artemisinin itself, induced significantly inhibited TNFα production. Artesunate, artemisone and DHA antagonized haemozoin-induced MMP-9 secretion by 25%, 24% and 50%, respectively. mRNA levels were also depressed by 14%, 20% and 27%, respectively, thus reflecting in part the effect observed on protein production. The derivatives significantly inhibited both TNFα-induced MMP-9 secretion and mRNA levels to a greater extent than haemozoin itself. Both haemozoin and TNFα increased NF-κB driven transcription by 11 and 7.7 fold, respectively. Artesunate, artemisone and DHA inhibited haemozoin-induced NF-κB driven transcription by 28%, 34%, and 49%, respectively. Similarly the derivatives, but not artemisinin, prevented TNFα-induced NF-κB driven transcription by 47-51%. The study indicates that artemisinins may attenuate the inflammatory potential of monocytes in vivo. Thus, in addition to direct anti-parasitic activities, the beneficial clinical effects of artemisinins for the treatment of malaria include the apparent ability to attenuate the inflammatory response, thus limiting the risk of progression to the more severe form of the disease, including the onset of cerebral malaria.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
361593
Artemisinin, 98%
C15H22O5
P0667
Parthenolide, ≥98% (HPLC)
C15H20O3