EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Formation of covalently closed circular DNA in Hep38.7-Tet cells, a tetracycline inducible hepatitis B virus expression cell line.
PMID 25150444
Abstract
Hepatitis B virus (HBV) covalently closed circular DNA (cccDNA) plays a central role in chronic HBV infection. However, analysis of the molecular mechanism of cccDNA formation is difficult because of the low efficiency in tissue cultured cells. In this study, we developed a more efficient cccDNA expression cell, Hep38.7-Tet, by subcloning from a tetracycline inducible HBV expression cell, HepAD38. Higher levels of cccDNA were produced in Hep38.7-Tet cells compared to HepAD38 cells. In Hep38.7-Tet cells, the cccDNA was detectable at six days after HBV induction. HBV e antigen (HBeAg) secretion was dependent upon cccDNA production. We screened chemical compounds using Hep38.7-Tet cells and HBeAg secretion as a marker. Most of the hit compounds have already been reported as anti-HBV compounds. These data suggested that Hep38.7-Tet cells will be powerful tools for analysis of the molecular mechanism of cccDNA formation/maintenance and development of novel therapeutic agents to control HBV infection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1103
Entecavir, ≥98% (HPLC)
C12H15N5O3