EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Identification of prometon, deisopropylprometon, and hydroxyprometon in groundwater by high resolution liquid chromatography/mass spectrometry.
PMID 25150740
Abstract
Prometon, a major soil sterilant, and its main transformation products, deisopropylprometon (N(2)-isopropyl-6-methoxy-1,3,5-triazine-2,4-diamine) and hydroxyprometon (4,6-bis(isopropylamino)-1,3,5-triazin-2-ol), were identified as the dominant triazine herbicides in groundwater samples from 51 locations in Colorado, USA, over a two-year time period. They were concentrated from water by solid phase extraction and detected using an ultrahigh pressure, liquid chromatography-quadrupole time of flight tandem mass spectrometry (UHPLC/QTOF-MS). The transformation products, deisopropylprometon and hydroxyprometon, were confirmed using MS-MS experiments. An original strategy was applied to form the degradation standards for deisopropylprometon and hydroxyprometon, which consisted of photo-degradation of prometon followed by MS-MS analysis. The concentration of prometon ranged from the detection limit of 3 ng·L(-1) to 87 ng·L(-1), hydroxyprometon ranged up to 50 ng·L(-1), and deisopropylprometon up to 100 ng·L(-1), with a frequency of detection of 80%, which was greater than the other triazines detected in the groundwater samples. A new ratio is proposed for prometon degradation called the "deisopropylprometon to prometon ratio" or the DIP ratio, as an indicator of prometon residence time in groundwater. Furthermore, these data suggest that prometon is more of an issue for groundwater contamination in urban areas rather than agricultural areas.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45635
Prometon, PESTANAL®, analytical standard
C10H19N5O
49086
Prometon, analytical standard
C10H19N5O