EMAIL THIS PAGE TO A FRIEND
American journal of obstetrics and gynecology
Treating morning sickness in the United States--changes in prescribing are needed.
PMID 25151184
Abstract
Presently, 97.7% of prescriptions for the treatment of nausea and vomiting in pregnancy in the United States are with medications not labeled for use in pregnancy, not indicated for nausea and vomiting in pregnancy, and not classified as safe in pregnancy by the Food and Drug Administration. The use of ondansetron for nausea and vomiting in pregnancy has increased from 50,000 monthly prescriptions in 2008 to 110,000 at the end of 2013, despite unresolved issues regarding fetal safety and Food and Drug Administration warnings about serious dysrhythmias. In April 2013, the Food and Drug Administration approved the combination of doxylamine and pyridoxine, specifically for nausea and vomiting in pregnancy symptoms. Now that a safe and effective drug is available in the United States, there is no reason for women to be exposed to a drug of unproven maternal and fetal safety.