EMAIL THIS PAGE TO A FRIEND
Cellular signalling
Ligand stimulation induces clathrin- and Rab5-dependent downregulation of the kinase-dead EphB6 receptor preceded by the disruption of EphB6-Hsp90 interaction.
PMID 25152371
Abstract
Ligand-induced internalisation and subsequent downregulation of receptor tyrosine kinases (RTKs) serve to determine biological outputs of their signalling. Intrinsically kinase-deficient RTKs control a variety of biological responses, however, the mechanism of their downregulation is not well understood and its analysis is focused exclusively on the ErbB3 receptor. The Eph group of RTKs is represented by the EphA and EphB subclasses. Each bears one kinase-inactive member, EphA10 and EphB6, respectively, suggesting an important role for these molecules in the Eph signalling network. While EphB6 effects on cell behaviour have been assessed, the mechanism of its downregulation remains elusive. Our work reveals that EphB6 and its kinase-active relative, and signalling partner, EphB4, are downregulated in a similar manner in response to their common ligand, ephrin-B2. Following stimulation, both receptors are internalised through clathrin-coated pits and are degraded in lysosomes. Their targeting for lysosomal degradation relies on the activity of an early endosome regulator, the Rab5 GTPase, as this process is inhibited in the presence of a Rab5 dominant-negative mutant. EphB6 also interacts with the Hsp90 chaperone and EphB6 downregulation is preceded by their rapid dissociation. Moreover, the inhibition of Hsp90 results in EphB6 degradation, mimicking its ligand-induced downregulation. These processes appear to rely on overlapping mechanisms, since Hsp90 inhibition does not significantly enhance ligand-induced EphB6 elimination. Taken together, our observations define a novel mechanism for intrinsically kinase-deficient RTK downregulation and support an intriguing model, where Hsp90 dissociation acts as a trigger for ligand-induced receptor removal.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E5029 Anti-EphB6 antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB2101093
Anti-HSP90AA1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2900334
Anti-HSP90AA1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2107614
Anti-HSP90AA1 antibody produced in rabbit, affinity isolated antibody
SAB2107615
Anti-HSP90AA1 antibody produced in rabbit, affinity isolated antibody
SAB4300541
Anti-HSP90AB1 (Ab-254) antibody produced in rabbit, affinity isolated antibody
SAB2701832
Anti-hsp90ab1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701806
Anti-hsp90ab1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV09016
Anti-HSP90AB1 antibody produced in rabbit, IgG fraction of antiserum
SAB2107344
Anti-HSP90AB1 antibody produced in rabbit, affinity isolated antibody
SAB4501462
Anti-HSP90B antibody produced in rabbit, affinity isolated antibody
SAB4501463
Anti-HSP90B antibody produced in rabbit, affinity isolated antibody
AV09018 Anti-HSPCB antibody produced in rabbit, IgG fraction of antiserum
SAB1305467
ANTI-HSPCB(N-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB3700701 Anti-Sheep IgG (Fc specific), highly cross adsorbed-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700700 Anti-Sheep IgG (Fc specific)-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700729 Anti-Sheep IgG (H+L), F(ab′)2 fragment-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700751 Anti-Sheep IgG (H+L), F(ab) fragment-Fluorescein antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700745 Anti-Sheep IgG (H+L), F(ab) fragment-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700687 Anti-Sheep IgG F(ab′)2, highly cross adsorbed-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700686 Anti-Sheep IgG F(ab′)2-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
EHU039481 MISSION® esiRNA, esiRNA human CLTC (esiRNA1)
EMU049451 MISSION® esiRNA, esiRNA targeting mouse Cltc (esiRNA1)
WH0002051M1
Monoclonal Anti-EPHB6 antibody produced in mouse, clone 5H6, purified immunoglobulin, buffered aqueous solution
SAB1403784
Monoclonal Anti-EPHB6 antibody produced in mouse, clone 5D8, purified immunoglobulin, buffered aqueous solution
SAB1408980
Monoclonal Anti-EPHB6 antibody produced in mouse, clone 3B6, purified immunoglobulin, buffered aqueous solution
SAB5300449
Monoclonal Anti-HSP90AA1 antibody produced in mouse, clone 5G5, purified immunoglobulin, buffered aqueous solution
SAB5300085 Monoclonal Anti-HSP90AB1 antibody produced in mouse, clone 5G4, ascites fluid
SAB5300086 Monoclonal Anti-HSP90AB1 antibody produced in mouse, clone 1D9, ascites fluid