PloS one
Meloxicam and buprenorphine treatment after ovarian transplantation does not affect estrous cyclicity and follicular integrity in aged CBA/J mice.
PMID 25153315
Abstract
Angiogenesis, the formation of new blood vessels, is important for the survival of ovarian transplants and the restoration of ovarian functions. Without angiogenesis, transplanted ovarian tissue becomes more susceptible to tissue damage and necrosis. Administration of analgesics for pain management has been shown to decrease angiogenesis, which can influence transplant success especially in aged animals. Aging and the effects of hypoxia after transplantation decrease reproductive viability of the ovarian transplant; therefore, it is important to understand the additional effects of analgesics on aged animal models. The present study investigated the effects of two analgesics, buprenorphine, an opiate, and meloxicam, a non-steroidal anti-inflammatory drug (NSAID), on the reproductive indicators related to estrous cyclicity and follicular integrity after ovarian transplantation of young ovaries into aged CBA/J mice. These aged females did not show any different reproductive responses when treated with either buprenorphine or meloxicam. No significant differences were observed in estrous cycle length, the onset of estrous cycling, the regularity of estrous cycles, and the proportion of viable follicles and total number of follicles per ovarian sample across treatment groups.