EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Nematicidal activity of natural ester compounds and their analogues against pine wood nematode, Bursaphelenchus xylophilus.
PMID 25153339
Abstract
In this study, we evaluated the nematicidal activity of natural ester compounds against the pine wood nematode, Bursaphelenchus xylophilus, to identify candidates for the development of novel, safe nematicides. We also tested the nematicidal activity of synthesized analogues of these ester compounds to determine the structure-activity relationship. Among 28 ester compounds tested, isobutyl 2-methylbutanoate, 3-methylbutyl 2-methylbutanoate, 3-methylbutyl tiglate, 3-methyl-2-butenyl 2-methylbutanoate, and pentyl 2-methylbutanoate showed strong nematicidal activity against the pine wood nematode at a 1 mg/mL concentration. The other ester compounds showed weak nematicidal activity. The LC50 values of 3-methylbutyl tiglate, isobutyl 2-methylbutanoate, 3-methylbutyl 2-methylbutanoate, 3-methyl-2-butenyl 2-methylbutanoate, and pentyl 2-methylbutanoate were 0.0218, 0.0284, 0.0326, 0.0402, and 0.0480 mg/mL, respectively. The ester compounds described herein merit further study as potential nematicides for pine wood nematode control.