EMAIL THIS PAGE TO A FRIEND
Molecular pharmaceutics
Self-immolative polycations as gene delivery vectors and prodrugs targeting polyamine metabolism in cancer.
PMID 25153488
Abstract
Polycations are explored as carriers to deliver therapeutic nucleic acids. Polycations are conventionally pharmacological inert with the sole function of delivering therapeutic cargo. This study reports synthesis of a self-immolative polycation (DSS-BEN) based on a polyamine analogue drug N(1),N(11)-bisethylnorspermine (BENSpm). The polycation was designed to function dually as a gene delivery carrier and a prodrug targeting dysregulated polyamine metabolism in cancer. Using a combination of NMR and HPLC, we confirm that the self-immolative polycation undergoes intracellular degradation into the parent drug BENSpm. The released BENSpm depletes cellular levels of spermidine and spermine and upregulates polyamine catabolic enzymes spermine/spermidine N(1)-acetyltransferase (SSAT) and spermine oxidase (SMO). The synthesized polycations form polyplexes with DNA and facilitate efficient transfection. Taking advantage of the ability of BENSpm to sensitize cancer cells to TNFα-induced apoptosis, we show that DSS-BEN enhances the cell killing activity of TNFα gene therapy. The reported findings validate DSS-BEN as a dual-function delivery system that can deliver a therapeutic gene and improve the outcome of gene therapy as a result of the intracellular degradation of DSS-BEN to BENSpm and the subsequent beneficial effect of BENSpm on dysregulated polyamine metabolism in cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
705284
1,1′-Carbonyldiimidazole solution, 0.4 M in methylene chloride
C7H6N4O
240117 1,6-Hexanediol, 99%
C6H14O2
H11807
1,6-Hexanediol, 97%
C6H14O2
D17408
1,7-Diaminoheptane, 98%
C7H18N2
717223
11-(1H-pyrrol-1-yl)undecane-1-thiol, 96%
C15H27NS
S7422
5-Sulfosalicylic acid dihydrate, BioXtra, ≥99.0%
C7H6O6S · 2H2O
S3147
5-Sulfosalicylic acid dihydrate, for electrophoresis, ≥99%
C7H6O6S · 2H2O
247006
5-Sulfosalicylic acid dihydrate, ACS reagent, ≥99%
C7H6O6S · 2H2O
S2130
5-Sulfosalicylic acid dihydrate, ReagentPlus®, ≥99%
C7H6O6S · 2H2O
86195
5-Sulfosalicylic acid dihydrate, purum p.a., ≥98.0% (T)
C7H6O6S · 2H2O
D2625
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
C12H12ClNO2S
39220
Dansyl chloride, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
C12H12ClNO2S
03641
Dansyl chloride, for HPLC derivatization, ≥99.0% (HPLC)
C12H12ClNO2S
M9671
Hexylene glycol, BioXtra, ≥99% (GC)
C6H14O2
ARK2151
Hexylene glycol, PharmaGrade, USP/NF, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
C6H14O2
68338
Hexylene glycol, BioUltra, ≥99.0% (GC)
C6H14O2
50797
Hexylene glycol, analytical standard
C6H14O2
68340
Hexylene glycol, puriss., ≥99.0% (GC)
C6H14O2
239550
Hexylene glycol, ≥99%
C6H14O2
112100
Hexylene glycol, 99%
C6H14O2
P0380
L-Proline, ReagentPlus®, ≥99% (HPLC)
C5H9NO2
P5607
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
C5H9NO2
P8865
L-Proline, PharmaGrade, Ajinomoto, EP, JP, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C5H9NO2
1568506
L-Proline, United States Pharmacopeia (USP) Reference Standard
C5H9NO2
81709
L-Proline, BioUltra, ≥99.5% (NT)
C5H9NO2
93693
L-Proline, certified reference material, TraceCERT®
C5H9NO2
W331902
L-Proline, 99%, FCC, FG
C5H9NO2
PHR1332
L-Proline, Pharmaceutical Secondary Standard; Certified Reference Material
C5H9NO2
S0266
Spermidine, BioReagent, for molecular biology, suitable for cell culture, ≥98%
C7H19N3
S2626
Spermidine, ≥99% (GC)
C7H19N3