EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Analysis of phthalates in milk and milk products by liquid chromatography coupled to quadrupole Orbitrap high-resolution mass spectrometry.
PMID 25155064
Abstract
A new analytical method was developed and validated for simultaneous analysis of 27 phthalates in milk and milk products. Response surface methodology was employed to optimize a quick, easy, cheap, effective, rugged, and safe (QuEChERS) sample preparation method. Ultrahigh-performance liquid chromatography and electrospray ionization quadrupole Orbitrap high-resolution mass spectrometry (UHPLC/ESI Q-Orbitrap) was used for the separation and detection of all the analytes. The method was validated by taking into consideration the guidelines specified in Commission Decision 2002/657/EC and 2007/19/EC. The extraction recoveries were in a range of 90.7% to 104.6%, with coefficient of variation <5.6%. The 27 compounds behave dynamic range in the 0.1-1000μgkg(-1) concentration, with correlation coefficient >0.99. The limits of detection for the analytes are in the range 0.32-2.6μgkg(-1). This method has been successfully applied on screening of phthalates in 96 commercial milk and milk product samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42438
Benzyl butyl phthalate, certified reference material, TraceCERT®
C19H20O4
36927
Benzyl butyl phthalate, analytical standard
C19H20O4
308501
Benzyl butyl phthalate, 98%
C19H20O4
442503
Benzyl butyl phthalate, analytical standard
C19H20O4
525197
Bis(2-ethylhexyl) adipate, 99%
C22H42O4
W519006
Bis(2-ethylhexyl) adipate, ≥99%
C22H42O4
02138
Bis(2-ethylhexyl) adipate, Selectophore, ≥99.0%
C22H42O4
02140
Bis(2-ethylhexyl) adipate, ≥97.0% (GC)
C22H42O4
442492
Bis(2-ethylhexyl) adipate, analytical standard
C22H42O4
67261
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®
C24H38O4
36735
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
C24H38O4
80030
Bis(2-ethylhexyl) phthalate, Selectophore
C24H38O4
753939
DBP, 98% (HPLC)
C64H36
88173
Di-n-octyl phthalate, certified reference material, TraceCERT®
C24H38O4
31301
Di-n-octyl phthalate, analytical standard
C24H38O4
80153
Di-n-octyl phthalate, ≥98.0% (GC)
C24H38O4
48560-U
Di-n-octyl phthalate, analytical standard
C24H38O4
309494
Dibutyl adipate, 96%
C14H26O4
D1010000
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
C16H22O4
80100
Dibutyl phthalate, Selectophore
C16H22O4
36736
Dibutyl phthalate, PESTANAL®, analytical standard
C16H22O4
36908
Dicyclohexyl phthalate, analytical standard
C20H26O4
306150
Dicyclohexyl phthalate, 99%
C20H26O4
454818
Diheptyl phthalate, 97%
C22H34O4
43540
Diisobutyl phthalate, certified reference material, TraceCERT®
C16H22O4
152641
Diisobutyl phthalate, 99%
C16H22O4
376663
Diisononyl phthalate, ester content ≥99 % (mixture of C9 isomers), technical grade
C26H42O4
80137
Diisopropyl phthalate, ≥97.0% (GC)
C14H18O4
D201154
Dioctyl phthalate, ≥99.5%
C24H38O4
525154
Dioctyl phthalate, 99%
C24H38O4