EMAIL THIS PAGE TO A FRIEND
Clinical and applied thrombosis/hemostasis : official journal of the International Academy of Clinical and Applied Thrombosis/Hemostasis
Red blood cell distribution width in patients with cryptogenic stroke.
PMID 25155500
Abstract
There is no information about a possible association of red blood cell distribution width (RDW) with cryptogenic stroke (CS). We aimed to analyze the association of RDW with CS. One hundred and sixty-three patients with CS were included along with 186 healthy controls. Fibrinogen, leukocytes, hemoglobin, and erythrocyte indices were evaluated. Patients showed higher RDW, leukocyte count, and body mass index (BMI) than controls (P < .05). No differences were observed in the erythrocyte indices or in glucose, cholesterol, and triglycerides levels (P > .05). When patients with anemia were excluded from the study (6 controls and 5 cases), the differences between cases and controls persisted (P = .005). Multivariate logistic regression revealed that, after adjusting for potential confounders (anemia, age > 40 years, gender, and fibrinogen >382 mg/dL, total cholesterol >240 mg/dL, and BMI > 28.7 kg/m(2)), RDW >14% was the only parameter that independently increased the risk of CS. The RDW >14% increased the risk of CS by 2.5-fold, irrespectively of anemia, inflammation, and lipidic profile.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
75018
1-Hexanethiol, analytical standard
C6H14S