EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Characterization of cross-protection by genetically modified live-attenuated Leishmania donovani parasites against Leishmania mexicana.
PMID 25156362
Abstract
Previously, we showed that genetically modified live-attenuated Leishmania donovani parasite cell lines (LdCen(-/-) and Ldp27(-/-)) induce a strong cellular immunity and provide protection against visceral leishmaniasis in mice. In this study, we explored the mechanism of cross-protection against cutaneous lesion-causing Leishmania mexicana. Upon challenge with wild-type L. mexicana, mice immunized either for short or long periods showed significant protection. Immunohistochemical analysis of ears from immunized/challenged mice exhibited significant influx of macrophages, as well as cells expressing MHC class II and inducible NO synthase, suggesting an induction of potent host-protective proinflammatory responses. In contrast, substantial inhibition of IL-10, IL-4, and IL-13 expression and the absence of degranulated mast cells and less influx of eosinophils within the ears of immunized/challenged mice suggested a controlled anti-inflammatory response. L. mexicana Ag-stimulated lymph node cell culture from the immunized/challenged mice revealed induction of IFN-γ secretion by the CD4 and CD8 T cells compared with non-immunized/challenged mice. We also observed suppression of Th2 cytokines in the culture supernatants of immunized/challenged lymph nodes compared with non-immunized/challenged mice. Adoptively transferred total T cells from immunized mice conferred strong protection in recipient mice against L. mexicana infection, suggesting that attenuated L. donovani can provide protection against heterologous L. mexicana parasites by induction of a strong T cell response. Furthermore, bone marrow-derived dendritic cells infected with LdCen(-/-) and Ldp27(-/-) parasites were capable of inducing a strong proinflammatory response leading to the proliferation of Th1 cells. These studies demonstrate the potential of live-attenuated L. donovani parasites as pan-Leishmania species vaccines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3701024 Anti-Mouse IgG (Fc specific)-Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701025 Anti-Mouse IgG (Fc specific)-Biotin antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701022 Anti-Mouse IgG (Fc specific)-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701023 Anti-Mouse IgG (Fc specific)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701020 Anti-Mouse IgG (Fc specific)-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701038 Anti-Mouse IgG (γ-chain specific)-Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701039 Anti-Mouse IgG (γ-chain specific)-Biotin antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701036 Anti-Mouse IgG (γ-chain specific)-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701037 Anti-Mouse IgG (γ-chain specific)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701034 Anti-Mouse IgG (γ-chain specific)-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701079 Anti-Mouse IgG (H+L) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701163 Anti-Mouse IgG (H+L), F(ab) fragment antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701166 Anti-Mouse IgG (H+L), F(ab) fragment-Biotin antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701164 Anti-Mouse IgG (H+L), F(ab) fragment-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701165 Anti-Mouse IgG (H+L), F(ab) fragment-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701162 Anti-Mouse IgG (H+L), F(ab) fragment-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701085 Anti-Mouse IgG (H+L)-Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3701087 Anti-Mouse IgG (H+L)-Biotin antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701088 Anti-Mouse IgG (H+L)-Biotin antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701081 Anti-Mouse IgG (H+L)-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701082 Anti-Mouse IgG (H+L)-Fluorescein antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701083
Anti-Mouse IgG (H+L)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701077 Anti-Mouse IgG (H+L)-Rhodamine antibody produced in rabbit, affinity isolated antibody, lyophilized powder
A4312 Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
A1902 Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
A2418 Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
B8520 Anti-Mouse IgG (whole molecule)−Biotin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
F9137 Anti-Mouse IgG (whole molecule)−FITC antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
T2402 Anti-Mouse IgG (whole molecule)−TRITC antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3700999 Anti-Mouse IgG F(ab′)2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution