EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. B
Compact and blinking-suppressed quantum dots for single-particle tracking in live cells.
PMID 25157589
Abstract
Quantum dots (QDs) offer distinct advantages over organic dyes and fluorescent proteins for biological imaging applications because of their brightness, photostability, and tunability. However, a major limitation is that single QDs emit fluorescent light in an intermittent on-and-off fashion called "blinking". Here we report the development of blinking-suppressed, relatively compact QDs that are able to maintain their favorable optical properties in aqueous solution. Specifically, we show that a linearly graded alloy shell can be grown on a small CdSe core via a precisely controlled layer-by-layer process, and that this graded shell leads to a dramatic suppression of QD blinking in both organic solvents and water. A substantial portion (>25%) of the resulting QDs does not blink (more than 99% of the time in the bright or "on" state). Theoretical modeling studies indicate that this type of linearly graded shell not only can minimize charge carrier access to surface traps but also can reduce lattice defects, both of which are believed to be responsible for carrier trapping and QD blinking. Further, we have evaluated the biological utility of blinking-suppressed QDs coated with polyethylene glycol (PEG)-based ligands and multidentate ligands. The results demonstrate that their optical properties are largely independent of surface coatings and solvating media, and that the blinking-suppressed QDs can provide continuous trajectories in live-cell receptor tracking studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T5625
(±)-α-Lipoic acid, synthetic, ≥99% (titration), powder
C8H14O2S2
T1395
(±)-α-Lipoic acid, BioReagent, cell culture tested, ≥99%
C8H14O2S2
62320
(±)-α-Lipoic acid, ≥98.0%
C8H14O2S2
87187
1-Tetradecene, analytical standard
C14H28
87189
1-Tetradecene, ≥97.0% (GC)
C14H28
T9805
1-Tetradecene, technical grade, 92%
C14H28
56454
1-Thioglycerol, PharmaGrade, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C3H8O2S
M1753
1-Thioglycerol, ≥97%
C3H8O2S
M6145
1-Thioglycerol, liquid, BioReagent, suitable for cell culture, ≥97% (titration)
C3H8O2S
M2172
1-Thioglycerol, BioXtra, ≥97%
C3H8O2S
88640
1-Thioglycerol, ≥99.0% (GC)
C3H8O2S
88642
1-Thioglycerol, dist., ≥98.0% (GC)
C3H8O2S
107700
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
C7H10N2
39405
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
C7H10N2
714844
4-(Dimethylamino)pyridine solution, 0.5 M in THF
C7H10N2
714720
4-(Dimethylamino)pyridine solution, 0.5 M in ethyl acetate
C7H10N2
700169
4-(Dimethylamino)pyridine, ChemDose tablets, Loading: 0.04 mmol tablet.
C7H10N2
M6035
4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt, powder
C16H17N2NaO9S
1368201
Alpha Lipoic Acid, United States Pharmacopeia (USP) Reference Standard
C8H14O2S2
525197
Bis(2-ethylhexyl) adipate, 99%
C22H42O4
W519006
Bis(2-ethylhexyl) adipate, ≥99%
C22H42O4
02138
Bis(2-ethylhexyl) adipate, Selectophore, ≥99.0%
C22H42O4
02140
Bis(2-ethylhexyl) adipate, ≥97.0% (GC)
C22H42O4
442492
Bis(2-ethylhexyl) adipate, analytical standard
C22H42O4
202894
Cadmium oxide, ≥99.99% trace metals basis
CdO
244783
Cadmium oxide, powder, 99.5% trace metals basis
CdO
633267
Dicyclohexylcarbodiimide solution, 60 wt. % in xylenes
C13H22N2
295299
Dimethyl ether, ≥99%
C2H6O
D201146
Dioctylamine, 97%
C16H35N
246654
Heptane, anhydrous, 99%
C7H16