EMAIL THIS PAGE TO A FRIEND
Infection, genetics and evolution : journal of molecular epidemiology and evolutionary genetics in infectious diseases
High diversity of beta-lactamases in the General Hospital Vienna verified by whole genome sequencing and statistical analysis.
PMID 25159028
Abstract
The detailed analysis of antibiotic resistance mechanisms is essential for understanding the underlying evolutionary processes, the implementation of appropriate intervention strategies and to guarantee efficient treatment options. In the present study, 110 β-lactam-resistant, clinical isolates of Enterobacteriaceae sampled in 2011 in one of Europe's largest hospitals, the General Hospital Vienna, were screened for the presence of 31 β-lactamase genes. Twenty of those isolates were selected for whole genome sequencing (WGS). In addition, the number of β-lactamase genes was estimated using biostatistical models. The carbapenemase genes blaKPC-2, blaKPC-3, and blaVIM-4 were identified in carbapenem-resistant and intermediate susceptible isolates, blaOXA-72 in an extended-spectrum β-lactamase (ESBL)-positive one. Furthermore, the observed high prevalence of the acquired blaDHA-1 and blaCMY AmpC β-lactamase genes (70%) in phenotypically AmpC-positive isolates is alarming due to their capability to become carbapenem-resistant upon changes in membrane permeability. The statistical analyses revealed that approximately 55% of all β-lactamase genes present in the General Hospital Vienna were detected by this study. In summary, this work gives a very detailed picture on the disseminated β-lactamases and other resistance genes in one of Europe's largest hospitals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1392454
Meropenem, United States Pharmacopeia (USP) Reference Standard
C17H25N3O5S · 3H2O