EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Cytohesin-associated scaffolding protein (CASP) is a substrate for granzyme B and ubiquitination.
PMID 25159843
Abstract
Natural killer (NK) cells are a sub-population of cytotoxic lymphocytes that can kill tumor or infected cells without prior exposure, by secreting the contents of preformed cytotoxic vesicles, containing perforin and granzymes, at the immune synapse. Cytohesin-associated scaffolding protein (CASP) is an adaptor molecule uniquely expressed in lymphocytes that forms complexes with both vesicle-initiating and sorting proteins, and has roles in NK cell migration, cytotoxicity, and cytokine secretion. In this study, we show that CASP contains a conserved granzyme B cleavage site that could modify its intracellular localization and interaction with sorting nexin 27. We also provide evidence that CASP is modified by ubiquitination. Both of these post-translational modifications could rapidly modify CASP function and highlight the dynamic regulatory mechanisms that direct its role in NK cell functions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA043598
Anti-CRTAP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA044150
Anti-CRTAP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105676
Anti-CRTAP, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1405681
Anti-CUX1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003277
Anti-CUX1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA003317
Anti-CUX1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105137
Anti-CUX1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1105737
Anti-CYTIP (286-298) antibody produced in rabbit, IgG fraction of antiserum
HPA007191
Anti-CYTIP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0010491M1
Monoclonal Anti-CRTAP antibody produced in mouse, clone 4D9, purified immunoglobulin, buffered aqueous solution
SAB1401528
Monoclonal Anti-CRTAP antibody produced in mouse, clone 2G5, purified immunoglobulin, buffered aqueous solution
WH0001523M1
Monoclonal Anti-CUTL1 antibody produced in mouse, clone 2A10, purified immunoglobulin, buffered aqueous solution
SAB1403715
Monoclonal Anti-CUTL1 antibody produced in mouse, clone 2D10, purified immunoglobulin, buffered aqueous solution
SAB4200043
Monoclonal Anti-Cytip antibody produced in rat, ~1.0 mg/mL, clone PDZ2F9, purified immunoglobulin, buffered aqueous solution