EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Rapid production of novel pre-microRNA agent hsa-mir-27b in Escherichia coli using recombinant RNA technology for functional studies in mammalian cells.
PMID 25161167
Abstract
Noncoding microRNAs (miRNAs or miRs) have been revealed as critical epigenetic factors in the regulation of various cellular processes, including drug metabolism and disposition. However, research on miRNA functions is limited to the use of synthetic RNA and recombinant DNA agents. Herein, we show that novel pre-miRNA-27b (miR-27b) agents can be biosynthesized in Escherichia coli using recombinant RNA technology, and recombinant transfer RNA (tRNA)/mir-27b chimera was readily purified to a high degree of homogeneity (>95%) using anion-exchange fast protein liquid chromatography. The tRNA-fusion miR-27b was revealed to be processed to mature miRNA miR-27b in human carcinoma LS-180 cells in a dose- and time-dependent manner. Moreover, recombinant tRNA/miR-27b agents were biologically active in reducing the mRNA and protein expression levels of cytochrome P450 3A4 (CYP3A4), which consequently led to lower midazolam 1'-hydroxylase activity. These findings demonstrate that pre-miRNA agents can be produced by recombinant RNA technology for functional studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
UC430
1′-Hydroxymidazolam
C18H13ClFN3O