PloS one
Spontaneous genomic alterations in a chimeric model of colorectal cancer enable metastasis and guide effective combinatorial therapy.
PMID 25162504
Abstract
Colon cancer is the second most common cause of cancer mortality in the Western world with metastasis commonly present at the time of diagnosis. Screening for propagation and metastatic behavior in a novel chimeric-mouse colon cancer model, driven by mutant p53 and β-Catenin, led to the identification of a unique, invasive adenocarcinoma. Comparison of the genome of this tumor, CB42, with genomes from non-propagating tumors by array CGH and sequencing revealed an amplicon on chromosome five containing CDK6 and CDK14, and a KRAS mutation, respectively. Single agent small molecule inhibition of either CDK6 or MEK, a kinase downstream of KRAS, led to tumor growth inhibition in vivo whereas combination therapy not only led to regression of the subcutaneous tumors, but also near complete inhibition of lung metastasis; thus, genomic analysis of this tumor led to effective, individualized treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100362
Anti-CCND1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100363
Anti-CCND1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2100364
Anti-CCND1 (ab3) antibody produced in rabbit, affinity isolated antibody
SAB1405510
Anti-CCND1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2106484
Anti-CCND1 antibody produced in rabbit, affinity isolated antibody
C5588 Anti-Cyclin D1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
H7157 Anti-Hepatocyte Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
H0652 Anti-Hepatocyte Growth Factor antibody produced in goat, IgG fraction of antiserum, lyophilized powder
H0540 Anti-Hepatocyte Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
H9786 Anti-Hepatocyte Growth Factor Receptor (c-Met) antibody produced in goat, affinity isolated antibody, lyophilized powder
H9911 Anti-Hepatocyte Growth Factor Receptor antibody produced in goat, lyophilized powder, affinity isolated antibody
SAB2101033
Anti-HGF (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101034
Anti-HGF (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2105571
Anti-HGF antibody produced in rabbit, affinity isolated antibody
HPA040360
Anti-HGF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA044088
Anti-HGF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410624
Anti-HGF antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300599
Anti-MET (Ab-1003) antibody produced in rabbit, affinity isolated antibody
SAB4300479
Anti-MET (Ab-1234) antibody produced in rabbit, affinity isolated antibody
SAB4300487
Anti-MET (Ab-1349) antibody produced in rabbit, affinity isolated antibody
SAB1406117
Anti-MET antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4501870
Anti-MET antibody produced in rabbit, affinity isolated antibody
HPA000451
Anti-MKI67 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001164
Anti-MKI67 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4700017 Monoclonal Anti-CCND1 antibody produced in mouse, clone CD1.1, purified immunoglobulin, buffered aqueous solution
SAB5300525
Monoclonal Anti-CCND1 antibody produced in mouse, clone 3D8, ascites fluid
C7464
Monoclonal Anti-Cyclin D1 antibody produced in mouse, clone DCS-6, ascites fluid
H1896 Monoclonal Anti-Hepatocyte Growth Factor antibody produced in mouse, clone 24612.111, purified immunoglobulin, lyophilized powder
H0665 Monoclonal Anti-Hepatocyte Growth Factor antibody produced in mouse, clone 24516, purified immunoglobulin, lyophilized powder
WH0004288M1
Monoclonal Anti-MKI67 antibody produced in mouse, clone 7B8, purified immunoglobulin, buffered aqueous solution