EMAIL THIS PAGE TO A FRIEND
Journal of microbiology (Seoul, Korea)
Characterization of a novel antigen of Mycobacterium tuberculosis K strain and its use in immunodiagnosis of tuberculosis.
PMID 25163841
Abstract
Mycobacterium tuberculosis-specific antigens would be of great value in developing immunodiagnostic tests for tuberculosis (TB), but regional differences in molecular types of the organism may result in antigenic variation, which in turn affects the outcome of the tests. For example, the Beijing strains of M. tuberculosis are prevalent in East Asia, and in particular, the K strain and related strains of the Beijing family, are most frequently isolated during school outbreaks of TB in South Korea. From comparison of genome sequences between M. tuberculosis K strain and the H37Rv strain, a non-Beijing type, we identified a K strain-specific gene, InsB, which has substantial homology with the ESAT-6-like proteins. This study was, therefore, initiated to characterize the InsB protein for its immunogenicity in mice and to confirm its expression in TB patients by detecting antibodies to the protein. The InsB gene was cloned from M. tuberculosis K strain and expressed in Escherichia coli. The recombinant InsB protein was used for immunization of mice. All mice showed strong antibody responses to the InsB protein, and splenocytes stimulated with InsB showed strong IFN-γ and IL-17 responses and a weak IL-2 response, all of which have been implicated in disease expression and used for the immunodiagnosis of TB. Serum samples from TB patients also showed significant antibody responses to the InsB protein as compared to healthy control samples. These results indicate that the InsB protein is an M. tuberculosis K-strain-specific antigen that could further improve the current immunodiagnostic methods, especially for the South Korean population.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3701270 Anti-Human IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701283 Anti-Human IgG (Fc specific), highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701282 Anti-Human IgG (Fc specific)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
A8419 Anti-Human IgG (γ-chain specific)−Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
SAB3701294 Anti-Human IgG (γ-chain specific)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701321 Anti-Human IgG (H+L), F(ab) fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701322 Anti-Human IgG (H+L), F(ab) fragment-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701360
Anti-Human IgG (H+L)-Peroxidase antibody produced in chicken, affinity isolated antibody, lyophilized powder
SAB3701359 Anti-Human IgG (H+L)-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3701358 Anti-Human IgG (H+L)-Peroxidase antibody produced in sheep, affinity isolated antibody, lyophilized powder
A8792 Anti-Human IgG (whole molecule)−Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3701242 Anti-Human IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701259 Anti-Human IgG F(ab′)2, highly cross adsorbed-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701257 Anti-Human IgG F(ab′)2-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701258 Anti-Human IgG F(ab′)2-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
D2779
Dimethyldioctadecylammonium bromide, ≥98% (TLC)
C38H80BrN
40163
Dimethyldioctadecylammonium bromide, Selectophore, ≥99.0%
C38H80BrN
40165
Dimethyldioctadecylammonium bromide, ≥98.0% (AT)
C38H80BrN