EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
Quantitative determination of metformin, glyburide and its metabolites in plasma and urine of pregnant patients by LC-MS/MS.
PMID 25164921
Abstract
This report describes the development and validation of an LC-MS/MS method for the quantitative determination of glyburide (GLB), its five metabolites (M1, M2a, M2b, M3 and M4) and metformin (MET) in plasma and urine of pregnant patients under treatment with a combination of the two medications. The extraction recovery of the analytes from plasma samples was 87-99%, and that from urine samples was 85-95%. The differences in retention times among the analytes and the wide range of the concentrations of the medications and their metabolites in plasma and urine patient samples required the development of three LC methods. The lower limit of quantitation (LLOQ) of the analytes in plasma samples was as follows: GLB, 1.02 ng/mL; its five metabolites, 0.100-0.113 ng/mL; and MET, 4.95 ng/mL. The LLOQ in urine samples was 0.0594 ng/mL for GLB, 0.984-1.02 ng/mL for its five metabolites and 30.0 µg/mL for MET. The relative deviation of this method was <14% for intra-day and inter-day assays in plasma and urine samples, and the accuracy was 86-114% in plasma, and 94-105% in urine. The method described in this report was successfully utilized for determining the concentrations of the two medications in patient plasma and urine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G117
Glipizide, solid
C21H27N5O4S
G0340000
Glipizide, European Pharmacopoeia (EP) Reference Standard
C21H27N5O4S