EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Selective inhibition of human solute carrier transporters by multikinase inhibitors.
PMID 25165131
Abstract
Solute carrier (SLC) transporters regulate the cellular influx and disposition of endogenous and xenobiotic compounds, including anticancer agents such as the multikinase inhibitors (MKIs). Recent evidence suggests that MKIs may also inhibit SLC-dependent transport of coadministered drugs, although present information on the relative susceptibilities of multiple SLC transporters is limited. This study evaluated 18 MKI drugs and metabolites as inhibitors of prototypic substrate uptake by 13 SLC transporters that were overexpressed in human embryonic kidney cells. Organic anion transporting polypeptides (OATPs) 1A2, 1B3, and 2B1, organic anion transporter 3 (OAT3), and organic cation transporter 1 (OCT1) were inhibited by most MKIs, whereas substrate uptake by OATP1B1, OAT1, 2, and 4, OCT2 and 3, and organic zwitterion/cation transporter 1 (OCTN1) was less susceptible to inhibition; OCTN2 was also inhibited by cediranib. In further studies, IC50 values were determined for the most effective MKIs, and erlotinib and cediranib were found to be potent competitive inhibitors of OATP2B1 (Ki = 41 nM) and OATP1A2 (Ki = 33 nM), respectively. From predictive approaches, several MKI-SLC interactions were found to be of potential in vivo significance.