EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
The cross-priming capacity and direct presentation potential of an autoantigen are separable and inversely related properties.
PMID 25165150
Abstract
We investigated whether a prevalent epitope of the β-cell-specific autoantigen islet-specific glucose-6-phosphatase catalytic subunit-related protein (IGRP206-214) reaches regional Ag-presentation pathways via unprocessed polypeptide chains, as free IGRP206-214 peptide or via preformed IGRP206-214/K(d) complexes. This was accomplished by expressing bacterial artificial chromosome transgenes encoding wild-type (stable) or ubiquitinated (unstable) forms of IGRP in IGRP-deficient NOD mice carrying MHC class I-deficient β-cells, dendritic cells, or B cells. We investigated the ability of the pancreatic lymph nodes of these mice to prime naive IGRP206-214-reactive CD8(+) T cells in vivo, either in response to spontaneous Ag shedding, or to synchronized forms of β-cell necrosis or apoptosis. When IGRP was made unstable by targeting it for proteasomal degradation within β-cells, the cross-priming, autoimmune-initiating potential of this autoantigen (designated autoantigenicity) was impaired. Yet at the same time, the direct presentation, CTL-targeting potential of IGRP (designated pathogenicity) was enhanced. The appearance of IGRP206-214 in regional Ag-presentation pathways was dissociated from transfer of IGRP206-214 or IGRP206-214/K(d) from β cells to dendritic cells. These results indicate that autoantigenicity and pathogenicity are separable and inversely related properties and suggest that pathogenic autoantigens, capable of efficiently priming CTLs while marking target cells for CTL-induced killing, may have a critical balance of these two properties.