EMAIL THIS PAGE TO A FRIEND
PLoS genetics
A cis-regulatory mutation of PDSS2 causes silky-feather in chickens.
PMID 25166907
Abstract
Silky-feather has been selected and fixed in some breeds due to its unique appearance. This phenotype is caused by a single recessive gene (hookless, h). Here we map the silky-feather locus to chromosome 3 by linkage analysis and subsequently fine-map it to an 18.9 kb interval using the identical by descent (IBD) method. Further analysis reveals that a C to G transversion located upstream of the prenyl (decaprenyl) diphosphate synthase, subunit 2 (PDSS2) gene is causing silky-feather. All silky-feather birds are homozygous for the G allele. The silky-feather mutation significantly decreases the expression of PDSS2 during feather development in vivo. Consistent with the regulatory effect, the C to G transversion is shown to remarkably reduce PDSS2 promoter activity in vitro. We report a new example of feather structure variation associated with a spontaneous mutation and provide new insight into the PDSS2 function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3773 Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal, clone HP-6017, purified from hybridoma cell culture
B5283 Anti-Rabbit IgG (γ-chain specific)−Biotin antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
R2655
Anti-Rabbit IgG, Native antibody, Mouse monoclonal, clone RabT-50, purified from hybridoma cell culture
G2904 Monoclonal Anti-Goat/Sheep IgG antibody produced in mouse, clone GT-34, ascites fluid
A8062 Monoclonal Anti-Goat/Sheep IgG–Alkaline Phosphatase antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous glycerol solution
B3148 Monoclonal Anti-Goat/Sheep IgG–Biotin antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous solution
F4891 Monoclonal Anti-Goat/Sheep IgG−FITC antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous solution
A9452 Monoclonal Anti-Goat/Sheep IgG–Peroxidase antibody produced in mouse, clone GT-34, purified immunoglobulin, lyophilized powder
I6135 Monoclonal Anti-Human IgG (Fab specific) antibody produced in mouse, clone GG-6, ascites fluid
I6260 Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone GG-7, ascites fluid
I6760 Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone HP-6017, ascites fluid
F5016 Monoclonal Anti-Human IgG (Fc specific)−FITC antibody produced in mouse, clone HP-6017, purified immunoglobulin, buffered aqueous solution
I5885 Monoclonal Anti-Human IgG (γ-chain specific) antibody produced in mouse, clone GG-5, ascites fluid
I9513 Monoclonal Anti-Human IgG2 (Fc specific) antibody produced in mouse, clone HP-6002, ascites fluid
I9888 Monoclonal Anti-Human IgG4 (pFc′ specific) antibody produced in mouse, clone HP-6023, ascites fluid
R1008 Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse, clone RG-96, ascites fluid
A2556 Monoclonal Anti-Rabbit IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in mouse, clone RG-96, purified immunoglobulin, buffered aqueous glycerol solution
R3155 Monoclonal Anti-Rabbit IgG, Native−Peroxidase antibody produced in mouse, clone RabT-50, purified from hybridoma cell culture
R0761 Monoclonal Anti-Rat IgG2a antibody produced in mouse, clone R2A-2, ascites fluid