EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Potassium-based algorithm allows correction for the hematocrit bias in quantitative analysis of caffeine and its major metabolite in dried blood spots.
PMID 25168119
Abstract
Although dried blood spot (DBS) sampling is increasingly receiving interest as a potential alternative to traditional blood sampling, the impact of hematocrit (Hct) on DBS results is limiting its final breakthrough in routine bioanalysis. To predict the Hct of a given DBS, potassium (K(+)) proved to be a reliable marker. The aim of this study was to evaluate whether application of an algorithm, based upon predicted Hct or K(+) concentrations as such, allowed correction for the Hct bias. Using validated LC-MS/MS methods, caffeine, chosen as a model compound, was determined in whole blood and corresponding DBS samples with a broad Hct range (0.18-0.47). A reference subset (n = 50) was used to generate an algorithm based on K(+) concentrations in DBS. Application of the developed algorithm on an independent test set (n = 50) alleviated the assay bias, especially at lower Hct values. Before correction, differences between DBS and whole blood concentrations ranged from -29.1 to 21.1%. The mean difference, as obtained by Bland-Altman comparison, was -6.6% (95% confidence interval (CI), -9.7 to -3.4%). After application of the algorithm, differences between corrected and whole blood concentrations lay between -19.9 and 13.9% with a mean difference of -2.1% (95% CI, -4.5 to 0.3%). The same algorithm was applied to a separate compound, paraxanthine, which was determined in 103 samples (Hct range, 0.17-0.47), yielding similar results. In conclusion, a K(+)-based algorithm allows correction for the Hct bias in the quantitative analysis of caffeine and its metabolite paraxanthine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5385
1,7-Dimethylxanthine, ~98%, solid
C7H8N4O2
93784
Caffeine, PharmaGrade, EP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
C8H10N4O2
C0750
Caffeine, powder, ReagentPlus®
C8H10N4O2
C8960
Caffeine, BioXtra
C8H10N4O2
C1778
Caffeine, Sigma Reference Standard, vial of 250 mg
C8H10N4O2
C0100000
Caffeine, European Pharmacopoeia (EP) Reference Standard
C8H10N4O2
C7731
Caffeine, meets USP testing specifications, anhydrous
C8H10N4O2
1085003
Caffeine, United States Pharmacopeia (USP) Reference Standard
C8H10N4O2
PHR1009
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10N4O2
56396
Caffeine, certified reference material, TraceCERT®
C8H10N4O2
W222402
Caffeine, anhydrous, 99%, FCC, FG
C8H10N4O2
05-0370
Caffeine, SAJ special grade, ≥98.5%
C8H10N4O2
Y0000787
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
C8H10N4O2
1086006
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
C8H10N4O2
PHR1095
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10N4O2
49897
Formate standard for IC, 1.000 g/L in H2O, analytical standard
CH2O2
06473
Formic acid solution, BioUltra, 1.0 M in H2O
CH2O2
41019
Melting point standard 235-237°C, analytical standard
C8H10N4O2
75035
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C8H10N4O2