EMAIL THIS PAGE TO A FRIEND
International journal of urology : official journal of the Japanese Urological Association
Contribution of stone size to chronic kidney disease in kidney stone formers.
PMID 25168662
Abstract
To determine whether stone burden correlates with the degree of chronic kidney disease in kidney stone formers. A total of 97 extracorporeal shockwave lithotripsy candidates aged 18 years and older were included. Size, number and location of the kidney stones, along with cumulative stone size, defined as the sum of diameters of all stones) were determined. Estimated glomerular filtration rate was determined using the Chronic Kidney Disease Epidemiology Collaboration cystatin C/creatinine equation, and chronic kidney disease was defined as estimated glomerular filtration rate <60 mL/min/1.73 m(2). In individuals with cumulative stone size <20 mm, estimated glomerular filtration rate significantly decreased when moving from the first (estimated glomerular filtration rate 75.5 ± 17.8 mL/min/1.73 m(2)) to the fourth (estimated glomerular filtration rate 56.4 ± 20.44 mL/min/1.73 m(2) ) quartile (P = 0.004). When patients with a cumulative stone size ≥ 20 mm were included, the observed association was rendered non-significant. In individuals with a cumulative stone size < 20 mm, each 1-mm increase in cumulative stone size was associated with a 20% increased risk of having chronic kidney disease. The relationship persisted even after adjustment for age, sex, body mass index, C-reactive protein, fasting plasma glucose, thyroid stimulating hormone, presence of microalbuminuria, history of renal calculi, history of extracorporeal shockwave lithotripsy, number and location of the stones (odds ratio 1.24, 95% confidence interval 1.02-1.52). The same was not observed for individuals with a cumulative stone size ≥ 20 mm. In kidney stone formers with a cumulative stone size up to 20 mm, estimated glomerular filtration rate linearly declines with increasing cumulative stone size. Additionally, cumulative stone size is an independent predictor of chronic kidney disease in this group of patients.