EMAIL THIS PAGE TO A FRIEND
Japanese journal of clinical oncology
A new human lung adenocarcinoma cell line harboring the EML4-ALK fusion gene.
PMID 25170107
Abstract
The echinoderm microtubule associated protein-like 4 (EML4)-anaplastic lymphoma kinase (ALK) fusion gene was identified in patients with non-small cell lung cancer. To the best of our knowledge, there are only three cell lines harboring the EML4-ALK fusion gene, which have contributed to the development of therapeutic strategies. Therefore, we tried to establish a new lung cancer cell line harboring EML4-ALK. A 61-year-old Japanese female presented with chest discomfort. She was diagnosed with left lung adenocarcinoma with T4N3M1 Stage IV. Although she was treated with chemotherapy, her disease progressed with massive pleural effusion. Because the EML4-ALK rearrangement was found in a biopsied specimen using fluorescence in situ hybridization, she was treated with crizotinib. She did well for 3 months. Tumor cells were obtained from the malignant pleural effusion before treatment with crizotinib. Cells continued to proliferate substantially for several weeks. The cell line was designated ABC-11. The EML4-ALK fusion protein and genes were identified in ABC-11 cells using fluorescence in situ hybridization and immunohistochemistry, respectively. ABC-11 cells were sensitive to crizotinib and next-generation ALK inhibitors (ceritinib and AP26113), as determined by a 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assay. Phosphorylated ALK protein and its downstream signaling were suppressed by treatment with crizotinib in western blotting. Furthermore, we could transplant ABC-11 cells subcutaneously into BALB/c nu/nu mice. We successfully established a new lung adenocarcinoma cell line harboring the EML4-ALK fusion gene. This cell line could contribute to future research of EML4-ALK-positive lung cancer both in vivo and in vitro.
Related Materials