EMAIL THIS PAGE TO A FRIEND
Atherosclerosis
Endothelial connexin 32 regulates tissue factor expression induced by inflammatory stimulation and direct cell-cell interaction with activated cells.
PMID 25171777
Abstract
Endothelial cell (EC) interacts with adjacent EC through gap junction, and abnormal expression or function of Cxs is associated with cardiovascular diseases. In patients with endothelial dysfunction, the up-regulation of tissue factor (TF) expression promotes the pathogenic activation of blood coagulation, however the relationship between gap junctions and TF expression in ECs remains uncharacterized. ECs express the gap junction (GJ) proteins connexin32 (Cx32), Cx37, Cx40 and Cx43. We investigated the role of endothelial gap junctions, particularly Cx32, in modulating TF expression during vascular inflammation. Human umbilical vein endothelial cells (HUVECs) were stimulated with tumor necrosis factor-α (TNF-α) and TF activity was assessed in the presence of GJ blockers and an inhibitory anti-Cx32 monoclonal antibody. Treatment with GJ blockers and anti-Cx32 monoclonal antibody enhanced the TNF-α-induced TF activity and mRNA expression in HUVECs. TNF-α-activated effector HUVECs or mouse MS-1 cells were co-cultured with non-stimulated acceptor HUVECs and TF expression in acceptor HUVECs was detected. Effector EC induced TF expression in adjacent acceptor HUVECs through direct cell-cell interaction. Cell-cell interaction induced TF expression was reduced by anti-intercellular adhesion molecule-1 (ICAM1) monoclonal antibody. Soluble ICAM1-Fc fusion protein promotes TF expression. GJ blockers and anti-Cx32 monoclonal antibody enhanced TF expression induced by cell-cell interaction and ICAM1-Fc treatment. Blockade of endothelial Cx32 increased TF expression induced by TNF-α stimulation and cell-cell interaction which was at least partly dependent upon ICAM1. These results suggest that direct Cx32-mediated interaction modulates TF expression in ECs during vascular inflammation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3595 Anti-Connexin-32 (106-124) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
C3470
Anti-Connexin-32 (265-279) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1405779
Anti-F3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA010663
Anti-GJB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV36600
Anti-GJB1 antibody produced in rabbit, IgG fraction of antiserum
SAB2104320
Anti-GJB1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
C6344
Monoclonal Anti-Connexin-32 antibody produced in mouse, clone CXN-32, ascites fluid, buffered aqueous solution
C8093
Monoclonal Anti-Connexin-43 antibody produced in mouse, clone CXN-6, ascites fluid
WH0002152M1
Monoclonal Anti-F3 antibody produced in mouse, clone 4G4, purified immunoglobulin, buffered aqueous solution
SAB1403798
Monoclonal Anti-F3 antibody produced in mouse, clone 4G4, ascites fluid
SAB1403864
Monoclonal Anti-GJB1 antibody produced in mouse, clone 2B9, purified immunoglobulin, buffered aqueous solution
SAB1402208
Monoclonal Anti-GJB1 antibody produced in mouse, clone 1F5, purified immunoglobulin, buffered aqueous solution
SAB1402940
Monoclonal Anti-TF antibody produced in mouse, clone 1C2, purified immunoglobulin, buffered aqueous solution
08393
Oleamide, analytical standard
C18H35NO
O2136
Oleamide, ≥99%
C18H35NO
O0140000
Plastic additive 20, European Pharmacopoeia (EP) Reference Standard
C18H35NO