EMAIL THIS PAGE TO A FRIEND
Immunobiology
Human endotoxin tolerance is associated with enrichment of the CD14+ CD16+ monocyte subset.
PMID 25172544
Abstract
Prior exposure to lipopolysaccharides (LPS) induces a state of cell resistance to subsequent LPS restimulation, known as endotoxin tolerance, mainly by repressing the expression of pro-inflammatory cytokines. We established an endotoxin tolerance model in human monocytes Endotoxin-tolerant cells showed a decrease in IκBα degradation and diminished expression of Tumor necrosis factor (TNF) (both messenger RNA [mRNA] and protein content). The myeloid differentiation factor 88 (MyD88)/MyD88 splice variant (MyD88s) ratio, an indirect way to test the Toll-like receptor 4 (TLR4) MyD88-dependent signaling cascade, did not change in endotoxin-tolerant cells when compared to LPS-stimulated or -unstimulated ones. Remarkably, cell population analysis indicated a significant increase of the CD14+ CD16+ subset only under the endotoxin-tolerant condition. Furthermore, endotoxin-tolerant cells produced higher amounts of C-X-C motif chemokine 10 (CXCL10), a typical MyD88-independent cytokine.