EMAIL THIS PAGE TO A FRIEND
International journal of food microbiology
Control of tyramine and histamine accumulation by lactic acid bacteria using bacteriocin forming lactococci.
PMID 25173915
Abstract
The aim of this study was to evaluate the competitive effects of three bacteriocin producing strains of Lactococcus lactis subsp. lactis against two aminobiogenic lactic acid bacteria, i.e. the tyramine producing strain Enterococcus faecalis EF37 and the histamine producing strain Streptococcus thermophilus PRI60, inoculated at different initial concentrations (from 2 to 6 log cfu/ml). The results showed that the three L. lactis subsp. lactis strains were able to produce bacteriocins: in particular, L. lactis subsp. lactis VR84 and EG46 produced, respectively, nisin Z and lacticin 481, while for the strains CG27 the bacteriocin has not been yet identified, even if its peptidic nature has been demonstrated. The co-culture of E. faecalis EF37 in combination with lactococci significantly reduced the growth potential of this aminobiogenic strain, both in terms of growth rate and maximum cell concentration, depending on the initial inoculum level of E. faecalis. Tyramine accumulation was strongly reduced when E. faecalis EF37 was inoculated at 2 log cfu/ml and, to a lesser extent, at 3 log cfu/ml, as a result of a lower cell load of the aminobiogenic strain. All the lactococci were more efficient in inhibiting streptococci in comparison with E. faecalis EF37; in particular, L. lactis subsp. lactis VR84 induced the death of S. thermophilus PRI60 and allowed the detection of histamine traces only at higher streptococci inoculum levels (5-6 log cfu/ml). The other two lactococcal strains did not show a lethal action against S. thermophilus PRI60, but were able to reduce its growth extent and histamine accumulation, even if L. lactis subsp. lactis EG46 was less effective when the initial streptococci concentration was 5 and 6 log cfu/ml. This preliminary study has clarified some aspects regarding the ratio between bacteriocinogenic strains and aminobiogenic strains with respect to the possibility to accumulate BA and has also showed that different bacteriocins can have different effects on BA production on the same strain. This knowledge is essentially aimed to use bacteriocinogenic lactococci as a predictable strategy against aminobiogenic bacteria present in cheese or other fermented foods.