EMAIL THIS PAGE TO A FRIEND
Oncology reports
Interaction of Muc4 and ErbB2 in a transgenic mouse model of gallbladder carcinoma: potential pathobiological implications.
PMID 25174601
Abstract
The molecular mechanism of gallbladder carcinogenesis and cancer growth remains unknown. BK5.erbB2 transgenic mice in which erbB2 is overexpressed and activated in the biliary epithelia develop adenocarcinoma of the gallbladder at a high incidence. Although it has been reported that erbB2 plays an important role in tumorigenesis, little is known about the involvement of its ligand(s). The expression level of Muc4, a potential functional ligand for erbB2, and its interaction with erbB2 in the gallbladder of BK5.erbB2 mice were determined. By immunohistochemistry and inxa0situ hybridization, both Muc4 mRNA and protein levels were strongly expressed in the cancerous epithelia of gallbladder from BK5.erbB2 mice. Also, in the hyperplastic (precancerous) epithelia, the protein levels were modestly expressed. Immunostaining with Muc4 (ASGP2) Ab overlapped with that with erbB2 Ab in the apical membranous components of the cancerous epithelia, indicating the co-localization of Muc4 and erbB2. Immunoprecipitation experiments revealed an interaction between Muc4 and erbB2 in the gallbladders. The interaction was associated with the hyperphosphorylation of erbB2, MAPK and Akt, and also with the overexpression of cyclooxygenase-2. However, in other organs that overexpressed erbB2 (trachea, esophagus and forestomach), Muc4 was expressed in only trace or modest amounts, and erbB2 was not hyperphosphorylated. Collectively, Muc4 is upregulated and interacts with erbB2 in gallbladders from BK5.erbB2 mice. It is likely that Muc4 plays an important role during gallbladder carcinogenesis and/or cancer growth by potentiating erbB2 signaling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2502079
Anti-ESR2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV31652
Anti-ESR2 antibody produced in rabbit, affinity isolated antibody
SAB2100713
Anti-ESR2 antibody produced in rabbit, affinity isolated antibody
SAB2105299
Anti-ESR2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
HPA005895 Anti-MUC4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700753
Anti-MUC4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900208
Anti-PTGES (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1406843
Anti-PTGES antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA045064
Anti-PTGES antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411671
Anti-PTGES antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
WH0002100M1
Monoclonal Anti-ESR2 antibody produced in mouse, clone 3F3, purified immunoglobulin, buffered aqueous solution
SAB2702145
Monoclonal Anti-ESR2 antibody produced in mouse
SAB2702146 Monoclonal Anti-ESR2 antibody produced in mouse
SAB2702147 Monoclonal Anti-ESR2 antibody produced in mouse
E1276 Monoclonal Anti-Estrogen Receptor β antibody produced in mouse, clone 9.88, ascites fluid
WH0004585M7
Monoclonal Anti-MUC4 antibody produced in mouse, clone 5B12, purified immunoglobulin, buffered aqueous solution
SAB1403031 Monoclonal Anti-PTGES antibody produced in mouse, clone 2B9, purified immunoglobulin, buffered aqueous solution