EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
Green synthesis, characterization and anti-inflammatory activity of silver nanoparticles using European black elderberry fruits extract.
PMID 25174985
Abstract
This research aimed at reporting the synthesis, characterization and evaluation of the anti-inflammatory effects of some new biomaterials based on silver nanoparticles and polyphenols rich natural extracts. A fast and eco-friendly extracellular biosynthesis of silver nanoparticles (AgNPs), using European black elderberry (Sambucus nigra - SN, Adoxaceae family) fruit extracts was developed. The phytosynthesized nanoparticles exhibited an absorbance peak at 426nm, characteristic for AgNPs and their sizes were ranged from 20 to 80nm. The anti-inflammatory properties of AgNPs were assessed in vitro on HaCaT cells exposed to UVB radiation, in vivo on acute inflammation model and in humans on psoriasis lesions. In vitro, our results demonstrated the anti-inflammatory effects of functionalized AgNPs by the decrease of cytokines production induced by UVB irradiation. In vivo, the pre-administration of AgNPs reduced the edema and cytokines levels in the paw tissues, early after the induction of inflammation. The present study also demonstrated the possible use of synthesized AgNPs for the treatment of psoriasis lesions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E5134
Ethylenediaminetetraacetic acid disodium salt dihydrate, for electrophoresis, for molecular biology, 99.0-101.0% (titration)
C10H14N2Na2O8 · 2H2O
E1644
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E6635
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, plant cell culture tested, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E5513
Ethylenediaminetetraacetic acid disodium salt dihydrate, for electrophoresis, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E4884
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
C10H14N2Na2O8 · 2H2O
ED2SS
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
C10H14N2Na2O8 · 2H2O
03677
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
C10H14N2Na2O8 · 2H2O
27285
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
C10H14N2Na2O8 · 2H2O
03685
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
C10H14N2Na2O8 · 2H2O
2854
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
C10H14N2Na2O8
E7889
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
C10H14N2Na2O8