EMAIL THIS PAGE TO A FRIEND
Oncology reports
hMSH2 expression is associated with paclitaxel resistance in ovarian carcinoma, and inhibition of hMSH2 expression inxa0vitro restores paclitaxel sensitivity.
PMID 25175513
Abstract
The objective of the present study was to investigate the association between paclitaxel resistance, gene copy number, and gene expression in ovarian carcinoma, and to restore paclitaxel sensitivity in a paclitaxel-resistant ovarian carcinoma cell line by using hMSH2-targeting siRNA. Paclitaxel-resistant ovarian carcinoma cell lines OC3/TAX300 and OC3/TAX50 and their parental cell lines were analyzed by comparative genomic hybridization, and the expression levels of hMSH2 in ovarian carcinoma cell lines and tissues were determined. An siRNA targeted to hMSH2 mRNA was used to transfect a paclitaxel-resistant cell line. We assessed the morphological features, proliferation, and susceptibility to apoptosis of the transfected cells after paclitaxel treatment. Chromosome 2p21xa0(gene locus of hMSH2) was amplified in OC3/TAX300 cells. hMSH2 was overexpressed in 93.9 and 47.6% of paclitaxel-treated and untreated ovarian carcinoma tissue samplesxa0(P=0.0001), respectively. hMSH2 was overexpressed in 93.3 and 54.2% of low-differentiated and moderate-to-highly differentiated ovarian carcinoma tissue samplesxa0(P=0.0008), respectively. hMSH2 expression was inhibited in the OC3/TAX300 cells transfected with hMSH2 siRNA. hMSH2 siRNA increased paclitaxel sensitivity, inhibited OC3/TAX300 cell proliferationxa0(G2/M arrest), and increased susceptibility to apoptosis. hMSH2 expression was upregulated in ovarian carcinoma cell lines and tissues after paclitaxel treatment. hMSH2 overexpression is related to paclitaxel resistance and poor prognosis. Inhibition of hMSH2 expression inxa0vitro restores paclitaxel sensitivity in paclitaxel‑resistant ovarian carcinoma cell lines and indicates a new direction in adjuvant therapy for ovarian carcinoma.